Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GGsTop
GREENER_CHOICESDP

Catalog No. 445210 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-5210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-5210 Supplier R&D Systems™ Supplier No. 4452/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of γ-glutamyl transpeptidase (GGT)

Selective, irreversible γ-glutamyl transpeptidase (GGT) inhibitor; displays no inhibitory effects on glutamine amidotransferases or asparagine synthetase. Suppresses oxidative stress by inhibiting GGT activation. Prevents ischemia/reperfusion-induced acute kidney injury in rats in vivo.

Chemical Identifiers

CAS 926281-37-0
Molecular Formula C13H18NO7P

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 331.26
Purity >98%
Molecular Weight (g/mol) 331.261
Chemical Name or Material GGsTop
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.