accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOONHOOHOH

CAS RN 931398-72-0

IUPAC Name: 2-[(1-benzyl-4-hydroxy-2-oxo-1,2-dihydroquinolin-3-yl)formamido]acetic acid
Synonyms: [(1-benzyl-4-hydroxy-2-oxoquinolin-3-yl)formamido]acetic acid
Molecular Weight (g/mol): 352.35
Molecular Formula: C19H16N2O5
InChi Key: CAOSCCRYLYQBES-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)C1=C(O)C2=CC=CC=C2N(CC2=CC=CC=C2)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results