accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOOOCH3H3CNHOOOCH3OOOHHOOOOHHO

CAS RN 932736-91-9

IUPAC Name: bis(methyl 2-[(4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl)cyclopropane-1-carboxylate); bis(oxalic acid)
Synonyms: bis(methyl 2-[(4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl)cyclopropane-1-carboxylate); bis(oxalic acid)
Molecular Weight (g/mol): 939.07
Molecular Formula: C52H62N2O14
InChi Key: IGJDSEGFEVKASG-UHFFFAOYNA-N
SMILES: OC(=O)C(O)=O.OC(=O)C(O)=O.COC(=O)C1(CC1CN1CCC(O)(CC1)C1=CC=CC=C1)C1=CC=C(C)C=C1.COC(=O)C1(CC1CN1CCC(O)(CC1)C1=CC=CC=C1)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)