Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ (±)-PPCC oxalate
GREENER_CHOICESDP

Catalog No. 387050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-705-0 50 mg
38-701-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-705-0 Supplier R&D Systems™ Supplier No. 3870/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective sigma (σ) agonist (σ1> σ2)

Selective sigma (σ) receptor ligand. Displays high affinity for σ1; also binds at σ2 sites (Ki = 1.5 nM and 50.8 nM respectively). Selective over a range of receptor types including dopaminergic and muscarinic receptors, DAT and SERT.

Chemical Identifiers

CAS 932736-91-9
Molecular Formula C52H62N2O14

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 469.53
Purity >99%
Molecular Weight (g/mol) 939.068
Chemical Name or Material (±)-PPCC oxalate
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.