CAS RN 934593-90-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ HIF-1 Inhibitor
The HIF-1 Inhibitor, also referenced under CAS 934593-90-5, controls the biological activity of HIF-1.
2
Tocris Bioscience™ LW 6
Malate dehydrogenase-2 (MDH2) inhibitor; also inhibits HIF-1α