Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LW 6
GREENER_CHOICESDP

Catalog No. 632250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-225-0 50 mg
63-221-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-225-0 Supplier Tocris Bioscience™ Supplier No. 6322/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Malate dehydrogenase-2 (MDH2) inhibitor; also inhibits HIF-1α

LW 6 is a malate dehydrogenase-2 (MDH2) inhibitor (IC50 = 6.3 μM). Also inhibits HIF-1α. Prevents accumulation of HIF-1α in Hep3B cells under hypoxic conditions (IC50 = 2.6 μM) with resulting decrease in expression of HIF-1α target genes VEGF and EPO. Inhibits proliferation of activated human T cells ex vivo.

Specifications

Chemical Name or Material Methyl 4-hydroxy-3-[[2-(4-tricyclo[3.3.1.13,7 ]dec-1-ylphenoxy)acetyl]amino]benzoate
CAS 934593-90-5
Quantity 50 mg
Target Dehydrogenase Inhibitors
Molecular Formula C26H29NO5
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.