accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 935693-62-2

IUPAC Name: N-(1-benzylpiperidin-4-yl)-6,7-dimethoxy-2-(4-methyl-1,4-diazepan-1-yl)quinazolin-4-amine
Synonyms: N-(1-benzylpiperidin-4-yl)-6,7-dimethoxy-2-(4-methyl-1,4-diazepan-1-yl)quinazolin-4-amine
Molecular Weight (g/mol): 490.65
Molecular Formula: C28H38N6O2
InChi Key: OSXFATOLZGZLSK-UHFFFAOYSA-N
SMILES: COC1=CC2=NC(=NC(NC3CCN(CC4=CC=CC=C4)CC3)=C2C=C1OC)N1CCCN(C)CC1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results