CAS RN 93602-66-5
IUPAC Name:
triethylazanium 6-(6-amino-9H-purin-9-yl)-7-hydroxy-2-sulfanylidene-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Synonyms:
TEAH 6-(6-aminopurin-9-yl)-7-hydroxy-2-sulfanylidene-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Molecular Weight (g/mol):
446.46
Molecular Formula:
C16H27N6O5PS
InChi Key:
OXIPZMKSNMRTIV-UHFFFAOYNA-N
SMILES:
CC[NH+](CC)CC.NC1=C2N=CN(C3OC4COP([O-])(=S)OC4C3O)C2=NC=N1
1
–
1
of
1
results
1
R&D Systems™ cAMPS-Sp, triethylammonium salt
CAS: 93602-66-5 Molecular Formula: C16H27N6O5PS