CAS RN 937171-63-6
CAS RN 937171-63-6
IUPAC Name:
(2R,3S)-3-{[(benzyloxy)(hydroxy)phosphoryl]oxy}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms:
(2R,3S)-3-{[benzyloxy(hydroxy)phosphoryl]oxy}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol):
511.47
Molecular Formula:
C26H26NO8P
InChi Key:
HOFDVXHILSPFNS-BXKMTCNYSA-N
SMILES:
C[C@H](OP(O)(=O)OCC1=CC=CC=C1)[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-D-Thr PO OBzl -OH -OH, Advanced ChemTech
CAS: 937171-63-6 Molecular Formula: C26H26NO8P Molecular Weight (g/mol): 511.47 MDL Number: MFCD07363481 InChI Key: HOFDVXHILSPFNS-BXKMTCNYSA-N IUPAC Name: (2R,3S)-3-{[(benzyloxy)(hydroxy)phosphoryl]oxy}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid SMILES: C[C@H](OP(O)(=O)OCC1=CC=CC=C1)[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O