accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNOHNCH3CH3HONNONNH2

CAS RN 937174-76-0

IUPAC Name: 4-[2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethyl-7-[(piperidin-3-yl)methoxy]-1H-imidazo[4,5-c]pyridin-4-yl]-2-methylbut-3-yn-2-ol
Synonyms: 4-[2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethyl-7-(piperidin-3-ylmethoxy)imidazo[4,5-c]pyridin-4-yl]-2-methylbut-3-yn-2-ol
Molecular Weight (g/mol): 425.49
Molecular Formula: C21H27N7O3
InChi Key: KGPGFQWBCSZGEL-UHFFFAOYNA-N
SMILES: CCN1C(=NC2=C(N=CC(OCC3CCCNC3)=C12)C#CC(C)(C)O)C1=NON=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)