accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOOSOONNH3C

CAS RN 93987-29-2

IUPAC Name: 2-methyl-1-(2-methyl-5-nitrobenzenesulfonyl)-1H-1,3-benzodiazole
Synonyms: 2-methyl-1-(2-methyl-5-nitrobenzenesulfonyl)-1,3-benzodiazole
Molecular Weight (g/mol): 331.35
Molecular Formula: C15H13N3O4S
InChi Key: ILVCPQPMRPHZSG-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=CC=C2N1S(=O)(=O)C1=C(C)C=CC(=C1)[N+]([O-])=O
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results