accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOSS

CAS RN 940-69-2

IUPAC Name: 5-(1,2-dithiolan-3-yl)pentanamide
Synonyms: lipoamide vitamin N thioctamide thioctic acid amide α-lipoic acid amide
Molecular Weight (g/mol): 205.33
Molecular Formula: C8H15NOS2
InChi Key: FCCDDURTIIUXBY-UHFFFAOYNA-N
SMILES: NC(=O)CCCCC1CCSS1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results