Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ (+/-)-a-Lipoamide, Phase Separated
Description
(±)-α-Lipoamide is a phase-separated condensate modifier. Reduces propensity of stress granule proteins to aggregate (EC50 = 20 μM in HeLa and iPSCs), and causes FUS stress granule proteins to return to the nucleus in vitro. Stimulates mitochondrial biogenesis in 3T3-L1 adipocytes. Prevents dieback of ALS patient-derived (FUS mutant) motor neuron axons in culture, and recovers motor defects in D. melanogaster expressing FUS mutants. Suppresses the effects of ALS-associated FUS mutations in vivo in different systems. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 1,2-Dithiolane-3-pentanamide |
| CAS | 940-69-2 |
| Quantity | 100 mg |
| Molecular Formula | C8H15NOS2 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?