CAS RN 941678-49-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Promotions Available
Ruxolitinib, Thermo Scientific™
Ruxolitinib, Thermo Scientific™
2
Tocris Bioscience™ Ruxolitinib
Potent and selective JAK1/JAK2 inhibitor; orally bioavailable