accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 944261-79-4

IUPAC Name: 5-(4-chlorophenyl)-N-(3,5-dimethoxyphenyl)furan-2-carboxamide
Synonyms: 5-(4-chlorophenyl)-N-(3,5-dimethoxyphenyl)furan-2-carboxamide
Molecular Weight (g/mol): 357.79
Molecular Formula: C19H16ClNO4
InChi Key: VHKBTPQDHDSBSP-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(NC(=O)C2=CC=C(O2)C2=CC=C(Cl)C=C2)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results