Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ A 803467
GREENER_CHOICESDP

Catalog No. 297650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-765-0 50 mg
29-761-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-765-0 Supplier R&D Systems™ Supplier No. 2976/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective NaV1.8 channel blocker

Selective blocker of NaV1.8 channels (IC50 values are 8, 2450, 6740, 7340 and 7380 nM for hNaV1.8, hNaV1.3, hNaV1.7, hNaV1.5 and hNaV1.2 channels respectively). Shows no significant activity against TRPV1, P2X2/3, CaV2.2 and KCNQ2/3 channels. Antinociceptive; potently attenuates mechanical allodynia in two models of neuropathic pain following i.p. administration.

Chemical Identifiers

CAS 944261-79-4
Molecular Formula C19H16ClNO4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 357.79
Purity >98%
Molecular Weight (g/mol) 357.79
Chemical Name or Material A 803467
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.