CAS RN 945725-50-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ TASP 0277308
Potent and selective S1P1 receptor antagonist; inhibits angiogenesis