accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrHNONN

CAS RN 946619-21-2

IUPAC Name: 4-(8-bromo-2-oxo-2,3-dihydro-1H-1,5-benzodiazepin-4-yl)pyridine-2-carbonitrile
Synonyms: 4-(7-bromo-4-oxo-3,5-dihydro-1,5-benzodiazepin-2-yl)pyridine-2-carbonitrile
Molecular Weight (g/mol): 341.17
Molecular Formula: C15H9BrN4O
InChi Key: KMKZCMKOSAKVGY-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)N=C(CC(=O)N2)C1=CC(=NC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)