CAS RN 947536-03-0
IUPAC Name:
N-{1-[3-(4-ethoxyphenyl)-4-oxo-3H,4H-pyrido[2,3-d]pyrimidin-2-yl]ethyl}-N-[(pyridin-3-yl)methyl]-2-[4-(trifluoromethoxy)phenyl]acetamide
Synonyms:
N-{1-[3-(4-ethoxyphenyl)-4-oxopyrido[2,3-d]pyrimidin-2-yl]ethyl}-N-(pyridin-3-ylmethyl)-2-[4-(trifluoromethoxy)phenyl]acetamide
Molecular Weight (g/mol):
603.6
Molecular Formula:
C32H28F3N5O4
InChi Key:
WQTKNBPCJKRYPA-UHFFFAOYNA-N
SMILES:
CCOC1=CC=C(C=C1)N1C(=O)C2=CC=CN=C2N=C1C(C)N(CC1=CN=CC=C1)C(=O)CC1=CC=C(OC(F)(F)F)C=C1
1
–
1
of
1
results
1
Tocris Bioscience™ (±)-AMG 487
CAS: 947536-03-0 Molecular Formula: C32H28F3N5O4