accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONONHOH

CAS RN 950762-95-5

IUPAC Name: N-hydroxy-1-[(4-methoxyphenyl)methyl]-1H-indole-6-carboxamide
Synonyms: N-hydroxy-1-[(4-methoxyphenyl)methyl]indole-6-carboxamide
Molecular Weight (g/mol): 296.33
Molecular Formula: C17H16N2O3
InChi Key: AJRGHIGYPXNABY-UHFFFAOYSA-N
SMILES: COC1=CC=C(CN2C=CC3=CC=C(C=C23)C(=O)NO)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)