Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PCI 34051
GREENER_CHOICESDP

Catalog No. 464350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-435-0 50 mg
46-431-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-435-0 Supplier R&D Systems™ Supplier No. 4643/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective HDAC8 inhibitor

Potent and selective histone deacetylase 8 (HDAC8) inhibitor (IC50 = 10 nM). Displays >200 fold selectivity over HDAC isoforms 1, 2, 3, 6 and 10. Induces apoptosis in cell lines derived from T cell lymphomas or leukemias.

Chemical Identifiers

CAS 950762-95-5
Molecular Formula C17H16N2O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 296.32
Molecular Weight (g/mol) 296.326
Chemical Name or Material PCI 34051
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.