CAS RN 96850-13-4
IUPAC Name:
3-(3,4-dichlorophenyl)-N-methyl-2,3-dihydro-1H-inden-1-amine hydrochloride
Synonyms:
3-(3,4-dichlorophenyl)-N-methyl-2,3-dihydro-1H-inden-1-amine hydrochloride
Molecular Weight (g/mol):
328.66
Molecular Formula:
C16H16Cl3N
InChi Key:
QICQDZXGZOVTEF-UHFFFAOYNA-N
SMILES:
Cl.CNC1CC(C2=CC=CC=C12)C1=CC(Cl)=C(Cl)C=C1
1
–
1
of
1
results
1
R&D Systems™ Indatraline hydrochloride
CAS: 96850-13-4 Molecular Formula: C16H16Cl3N