accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 96865-83-7

IUPAC Name: 2-[4-(2,6-dioxo-1,3-dipropyl-2,3,6,7-tetrahydro-1H-purin-8-yl)phenoxy]acetic acid
Synonyms: 4-(2,6-dioxo-1,3-dipropyl-7H-purin-8-yl)phenoxyacetic acid
Molecular Weight (g/mol): 386.41
Molecular Formula: C19H22N4O5
InChi Key: QTMMGCYGCFXBFI-UHFFFAOYSA-N
SMILES: CCCN1C2=C(NC(=N2)C2=CC=C(OCC(O)=O)C=C2)C(=O)N(CCC)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results