accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3ClH2N

CAS RN 97-50-7

IUPAC Name: 5-chloro-2,4-dimethoxyaniline
Synonyms: aniline,2,4-dimethoxy-5-chloro
Molecular Weight (g/mol): 187.62
Molecular Formula: C8H10ClNO2
InChi Key: OLCMNCWEUMBNIS-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(Cl)C=C1N
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)