accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 98048-99-8

CH3NH2POOHHO

CAS RN 98048-99-8

IUPAC Name: (1-aminopropyl)phosphonic acid
Synonyms: 1-aminopropylphosphonic acid
Molecular Weight (g/mol): 139.09
Molecular Formula: C3H10NO3P
InChi Key: DELJNDWGTWHHFA-UHFFFAOYNA-N
SMILES: CCC(N)P(O)(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results