CAS RN 99095-10-0
IUPAC Name:
4-{4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl}-4-azatricyclo[5.2.1.0²,⁶]decane-3,5-dione hydrochloride
Synonyms:
4-{4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl}-4-azatricyclo[5.2.1.0²,⁶]decane-3,5-dione hydrochloride
Molecular Weight (g/mol):
419.95
Molecular Formula:
C21H30ClN5O2
InChi Key:
ACVFJYKNBOHIMH-UHFFFAOYNA-N
SMILES:
Cl.O=C1C2C3CCC(C3)C2C(=O)N1CCCCN1CCN(CC1)C1=NC=CC=N1
1
–
1
of
1
results
1
R&D Systems™ Tandospirone hydrochloride
CAS: 99095-10-0 Molecular Formula: C21H30ClN5O2