CAS RN 99247-33-3
IUPAC Name:
sodium 4-(dipropylcarbamoyl)-4-(phenylformamido)butanoate
Synonyms:
sodium 4-(dipropylcarbamoyl)-4-(phenylformamido)butanoate
Molecular Weight (g/mol):
356.4
Molecular Formula:
C18H25N2NaO4
InChi Key:
UFMWGCINVOIJSO-UHFFFAOYNA-M
SMILES:
[Na+].CCCN(CCC)C(=O)C(CCC([O-])=O)NC(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
R&D Systems™ Proglumide sodium salt
CAS: 99247-33-3 Molecular Formula: C18H25N2NaO4