CAS RN 99636-32-5
IUPAC Name:
1-methoxypropan-2-amine
Synonyms:
1-methoxypropan-2-amine
Molecular Weight (g/mol):
89.14
Molecular Formula:
C4H11NO
InChi Key:
NXMXETCTWNXSFG-UHFFFAOYNA-N
SMILES:
COCC(C)N
1
–
2
of
2
results
1
Sigma Aldrich (S)-1-Methoxy-2-Propylamine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.