Buy 3 Sure-Vue Test Kits for the Price of 2
For a limited time, buy three Fisher Healthcare™ Sure-Vue™ Signature Cryptosporidium/Giardia Diagnostic Test/Control Kits for the price of two.
If you’ve already purchased your kits, please fill out the form on this page and upload your proof of purchase to receive your additional kit. If you’ve not yet made your purchase but are interested in redeeming this great offer, contact your sales representative to learn more.
Features and Benefits
Sure-Vue Signature Cryptosporidium/Giardia Diagnostic Test/Control Kits provide fast, accurate results for the two most common intestinal parasite infections — all in a single test.
- One test that both detects and differentiates between cryptosporidium and giardia
- Proven performance with high sensitivity (cryptosporidium: 97%/giardia: 100%) and specificity (cryptosporidium: 99.5%/giardia: 99.1%)
- Improved turnaround time — test results in approximately 10 minutes
Due to product restrictions, we cannot sell these products online. Contact your local sales representative for more information or to place an order.
You can also find out more about these products by contacting Customer Service by phone at 1-800-640-0640 or via email at CS.WebEmail@thermofisher.com.
Restrictions
Offer valid until March 31, 2018.
Customer acknowledges that this offer may include a discount or other price reduction that must be properly and accurately accounted for and reported by customer in accordance with all federal and state laws, including without limitation the federal anti-kickback law (42 U.S.C. § 1320a-7b(b)(3)(A)) and regulations thereunder (42 C.F.R. §1001.952(h)).