Diasorin LIAISON NES System
One Swab. Four Answers. Same Visit.
In high-volume environments where respiratory symptoms overlap, result clarity can help reduce uncertainty and keep your patient management moving forward.
Enable accurate, same-visit decisions with the next-generation Diasorin system. The LIAISON NES™ molecular system delivers real-time RT-PCR detection of influenza A, influenza B, RSV, and SARS-CoV-2 in about 15 minutes, supporting confident treatment, isolation accuracy, and antibiotic stewardship.
Enable Faster Personalized Care
Simplify end-to-end molecular testing in your acute care setting. The LIAISON NES system delivers a complete sample-to-result molecular solution that integrates seamlessly into your workflows.
It supports:
- Dry swab sampling and room-temperature reagent stability
- Lab-grade accuracy that clinicians can trust
- CLIA-waived molecular testing at the point of care
- Guided automation that simplifies testing workflows
- Streamlined onboarding with minimal training requirements
- Centralized visibility through connected oversight
Get diagnostic confidence with operational simplicity.
Accelerate Respiratory Molecular Testing with the LIAISON NES™ System
When Every Minute Counts, Clear Results Matter
Hands-On Time Is for Patients
A simple three-step workflow reduces hands-on time to under one minute—so you can spend more time with patients and less time handling samples.
For more information, contact your Fisher Healthcare representative.
Discover the LIAISON NES Solution
Explore the LIAISON NES system and assay solutions to learn more about product features and capabilities.
Explore More
Learn More About the Diasorin Point-of-Care Device
Request More Information
Interested in bringing Diasorin LIAISON NES™ rapid 4-in-1 respiratory molecular testing to your acute care setting? Connect with a Fisher Healthcare representative to discuss implementation, training, and connectivity.