UserName
Viewing profile as user:
Fisherbrand Histology Slide Comparison Guide
Fisherbrand Histology Slide Comparison Guide
Choose the Right Slide for Your Histology Application
Your laboratory results rely on the selecting the right product for your testing measures and applications.
When you choose Fisherbrand slides for your histology applications, you have access to an unmatched breadth of options and flexibility to meet your needs. Made from Swiss white glass, Fisherbrand microscope slides are selected for uniform quality and reliable clarity, with precision-ground edges and consistent dimensional tolerances. Their excellent adhesion and hydrophilic properties offer dependable, high-quality, consistent performance with instrumentation. In addition, each slide meets stringent quality control standards to ensure that your lab operations and service levels can be reliably maintained.
Adhesion Slides Comparison
Superfrost Plus Slides
Adhesion workhorse slide with excellent hydrophilic and adhesive properties, electrostatically charged, trusted reliability with instrumentation.
Benefits: Holds tissue without need for expensive coating steps, virtually eliminates tissue loss during staining and saves hours of slide preparation.
Superfrost Plus Slides
Superfrost Plus Gold
Advanced adhesion coating attracts and then chemically bonds fresh or frozen tissue.
Benefits: For hard-to-hold tissue samples like bone, brain and breast that are larger than 5 microns.
Superfrost Plus Gold
Superfrost Excell
Enhanced performance characteristics over Superfrost Plus.
Benefits: Particularly effective in HIER methods that require high pH antigen retrieval solutions, including EDTA.
Superfrost Excell
Polysine
Adhesive slide with similar properties to Silane or Poly-L-Lysine coated slides.
Benefits: Attracts frozen and paraffin embedded sections, cytocentrifuge preps and cytology smears.
Polysine
Colormark Plus
Designed for automated etched labeling machines, Superfrost Plus performance characteristics.
Benefits: Provides excellent etched labeling visibility in a range of slide tab colors.
Colormark Plus
Inkjet Plus
Designed for inkjet printing instruments, Superfrost Plus performance characteristics.
Benefits: Chemical resistant inks provide clearly visible text and bar codes on the slide tab.
Inkjet Plus