Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Colormark™ Standard Microscope Slide

Catalog No. 22038329
Click to view available options
:
Lavender w/Black, Pack of 144
This item is not returnable. View return policy
This item is not returnable. View return policy

For use with SHUR/Mark™ labeling system

 

  • Colormark slides are designed for slide etching instruments
  • A contrasting color is printed on the back of the Superfrost end so that the patient or specimen information etched through the painted end on the front is clearly visible
  • Lavendar w/Black

 

Specifications

Color Lavender, Black
Coating Non-adhesion
Quantity 144 Pk., 10 Pk./Cs.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.