Pharmacogenomics
Simply Your Pharmacogenomics Research
Go from sample to genotyping results with ease with Applied Biosystems™ pharmacogenomics (PGx) products. Count on an innovative portfolio of genetic analysis tools to help you develop essential tests for analyzing the genetic variations involved in responses to medication.
These include low- and high-throughput systems that can be customized to meet your specific pharmacogenomics research needs. Choose from two main solutions:
- Real-time PCR for investigating fewer than 120 targets
- Next-generation sequencing for analyzing large numbers of genetic markers (120+)*
For research use only.
*Contact your Fisher Healthcare account manager to learn more.
- TaqMan Assays
- Connect With Us
A Flexible, Time-Saving Solution
Enable ReliablePGx Decision-Making
Enable flexible, high-throughput screening and cut costs with these state-of-the art genetic analysis solutions. When working together, the Applied Biosystems™ QuantStudio™ 12K Flex Real-Time PCR System, Applied Biosystems™ OpenArray™ Plates, and qualified Applied Biosystems™ TaqMan® and TrueMark™ Assays deliver results quickly for a simpler, more efficient workflow.
Experimental Workflows
Streamline your PGx workflow withTrueMark PGx SNP + CNV Real-Time PCR panels designed to simultaneously detect single-nucleotide polymorphisms (SNPs) and copy number variations (CNVs).
They help you maximize your data analysis efficiency by providing and analyzing genotyping results in one run. The process completes in as little as 5 hours* so you can swiftly begin another run within a typical 8-hour shift.
Key Benefits
- Improved efficiency—Go from sample to genotyping results in a single-run workflow with integrated data analysis
- Accurate and relevant answers, your way—Employ guideline-driven fixed panel content in multiple qPCR formats using customization capabilities
Featured Products
Catalog Number A40003215
Catalog Number A65342
Catalog Number 4472380
Catalog Number 501527922
Accelerate Your Validation Process
Reach out to an account representative to request dedicated analytical validation consulting and technical support services to help your research lab implement, operate, and scale more efficiently.