Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TrueMark™ PGx SNP + CNV 64 Panel, 384-well plate

Catalog No. A40003215
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Plates (60 Samples)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40003215 10 Plates (60 Samples)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A40003215 Supplier Applied Biosystems™ Supplier No. A40003215
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The TrueMark PGx SNP + CNV 64 Panel is a pre-spotted 384-well PCR plate that is easy-to-use for low to mid-throughput pharmacogenomics research sample testing applications. The panel includes 56 SNP assays and 2 CNV assays.

This TrueMark™ PGx (Pharmacogenomics) SNP + CNV 64 Panel is offered in pre-spotted 384-well plates, an efficient, easy-to-use format for pharmacogenomics applications. This panel contains 56 TaqMan® DME and SNP genotyping assays in singlicate and two CNV assays in quadruplicates allowing the testing of up to six DNA samples per 384-well plate. The assays were developed to detect polymorphisms in genes encoding drug metabolism enzymes (DMEs) and associated transport proteins. This panel provides coverage of essential, commonly studied markers within CYP2D6, CYP2C19, CYP2C9, DPYD and other important DME and clinical research genes.

Each TaqMan Drug Metabolism and SNP genotyping assay in the panel contains two allele-specific probes and a primer pair to detect the specific SNP target. Both the probes and primers uniquely align within the genome, enabling the TaqMan genotyping technology to provide superior specificity. It is this specificity that allows these assays to detect targets residing in highly homologous gene families that may include pseudogenes. Each copy number assay is combined with a TERT reference assay in a single well to form a duplex assay that is designed to detect copy number variation (CNV) such as deletion, duplication or hybrid alleles.

TaqMan Drug Metabolism and genotyping and CNV assays were developed using a high level of bioinformatics and wet-lab stringency. All assays have passed performance tests involving 180 unique DNA samples from four different populations.

The plate is pre-formatted and inventoried, cost-effective, convenient, and easy-to-use, without the need for expensive robotics. Results are reproducible and consistent across samples, studies, and labs—for the same data quality from plate-to-plate and lot-to-lot, even with different operators.

For fast delivery, all assays in this collection have been manufactured and placed into inventory and are ready to ship at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Detection Method TaqMan
Form Dried Down
GC-Rich PCR Performance Medium
PCR Method qPCR
Reaction Speed Standard
Technique qPCR
Label or Dye FAM + VIC
No. of Tests 6 Samples per 384-well Plate (60 Samples)
Quantity 10 Plates (60 Samples)
Shipping Condition Room Temperature
Species Human
Target Organism Class Human
For Use With (Application) Genotyping, Copy Number Variation Analysis, Real Time PCR (qPCR)
No. of Reactions 64 Reactions/Sample
Sample Type Buccal swabs and EDTA whole blood
Size (Final Product) 10 Plates
Certification Compliance ISO 9001
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.