Crucibles
Fisherbrand™ PTFE Crucibles
Withstand temperatures to 280°C (536°F).
Fisherbrand™ High-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F)
Fisherbrand™ Alumina Crucibles
Made up of 99.7% Alumina
Fisherbrand™ Nickel Crucibles and Covers
Fisherbrand Nickel Crucibles can withstand temperatures up to 800°C (1472°F)
PYREX™ Gooch Type Filtering Crucibles Available on GSA/VA Contract for Federal Government customers only.
For drying precipitates to constant weight at 110°C (230°F)
Fisherbrand™ Porcelain Casserole Crucibles
For use in laboratory studies
DWK Life Sciences Kimble™ KIMAX™ Gooch Filtering Crucibles Available on GSA/VA Contract for Federal Government customers only.
Dry precipitates to constant weight at 110°C (230°F)
Alfa Aesar™ Graphite Fusion Crucible
Manufactured from graphite materials that provide exceptional thermal uniformity and exactness to dimensional specifications.
Fisherbrand™ Porcelain Bitumen Filtering Crucible
Filtration device for general laboratory use
QSI Fused-Quartz Crucibles
Withstand temperatures up to 1250°C (2282°F)
Fisherbrand™ Zirconium Crucibles with Cover
Fisherbrand Zirconium Crucibles with Cover can withstand about 100 two- to five- minute fusions between 460°C (860°F) and 900°C (1652°F)
Fisherbrand™ Wide-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F)
Fisherbrand™ Nickel Crucibles and Covers
Fisherbrand Nickel Crucibles can withstand temperatures up to 800°C (1472°F)
Saint-Gobain ALUNDUM™ Filtering Crucibles
For melting metals and vaporizing liquids
CoorsTek™ High-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
CoorsTek™ Porcelain Desiccator Plates for Crucibles
With 30mm diameter holes for crucibles
QSI Fused Quartz Crucible Covers
For 08-072 Series Crucibles
Synthware™ Low Form Gooch Crucibles
With sealed-in fritted disc of shown porosity
Fisherbrand™ Porcelain Crucible Covers
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles
CoorsTek™ Porcelain Gooch Filtering Crucibles
Glazed inside and out except for outside bottom surface
Fisherbrand™ Porcelain Gooch Filtering Crucibles
For total suspended solids determinations
Fisherbrand™ Porcelain 90mL Dish Crucible
For laboratory and industrial purpose
Fisherbrand™ Porcelain Rose Crucible
For use in very high temperatures
Fisherbrand™ Porcelain Flat Capsule Crucibles
For use in laboratories of educational institutions, scientific development and industrial research, chemical, fertilizer, petro-chemical establishments
Fisherbrand™ Porcelain Capsule-Form Crucibles
Glazed inside and out except for rim
Plasticoid Walter Crucible Holder
Rubber with removable glass stem ground at 45° angle
  spinner