Cell Dividers, Inserts, Scrapers and Utensils
Corning™ Transwell™ Multiple Well Plate with Permeable Polyester Membrane Inserts Available on GSA/VA Contract for Federal Government customers only.
Featuring a microscopically transparent polyester membrane that is TC-treated for optimal cell attachment and growth. Corning™ Transwell™ Multiwell Plate with Polyester Membrane Inserts provide better visibility under phase contrast microscopy and allow assessment of cell viability and monolayer formation.
Falcon™ Round-Bottom Polystyrene Test Tubes with Cell Strainer Snap Cap Available on GSA/VA Contract for Federal Government customers only.
Designed for flow cytometry applications. Corning™ Falcon™ Test Tube with Cell Strainer Snap Cap offer a convenient solution to prepare laboratory samples.
Fisherbrand™ Cell Scrapers
Easily harvest cells from tissue culture flasks
Falcon™ Standard Tissue Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Constructed of flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Corning Falcon™ Standard Tissue Culture Dishes works with most attachment-dependent cell lines.
Corning™ Cell Lifter Available on GSA/VA Contract for Federal Government customers only.
Designed with a wide beveled edge for cell culture dishes. Corning™ Cell Lifters are an ideal tool for harvesting stem cells and colonies while maintaining high viability.
Falcon™ Cell Strainers Available on GSA/VA Contract for Federal Government customers only.
Rapidly isolates primary cells to consistently obtain a uniform single-cell suspension from tissues. Corning Falcon™ Cell Strainers are easy to use and ideal for use with stem cells and primary cells.
Falcon™ Cell Scrapers Available on GSA/VA Contract for Federal Government customers only.
Designed to provide maximum accessibility to growth surfaces. Corning™ Falcon™ Cell Scrapers provide superior scraping ability with dishes, plates, and flasks.
Fisherbrand™ Cell Lifters
Disposable lifters quickly remove cell layers
PYREX™ Cloning Cylinders Available on GSA/VA Contract for Federal Government customers only.
Create identical cells by surrounding a single cell or group of cells
Fisherbrand™ Sterile Cell Strainers
Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.
Corning™ Cell Scrapers Available on GSA/VA Contract for Federal Government customers only.
Angled swivel blade minimizes cell damage and ensures even contact with growth area. Corning™ Cell Scrapers are ideal for harvesting cells.
Corning™ BioCoat™ Matrigel™ Invasion Chamber with Corning™ Matrigel Matrix Available on GSA/VA Contract for Federal Government customers only.
Provide rapid and reproducible quantification of cell invasion in vitro. Corning™ BioCoat™ Matrigel™ Invasion Chamber with Corning Matrigel Matrix™ cell culture inserts are pre-coated with extracellular matrix proteings for applicatins requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and differentiation.
MilliporeSigma™ Millicell™ Cell Culture Insert Plates Available on GSA/VA Contract for Federal Government customers only.
Designed to support cell growth and differentiation of endothelial and epithelial cell lines
Falcon™ Cell Culture Inserts Available on GSA/VA Contract for Federal Government customers only.
Use with 24-well plates. Corning™ Falcon™ Cell Culture Inserts (cell culture on permeable supports) results in greater morphological and/or physiological differentiation of many cell types.
Corning™ TC-Treated Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Tissue-culture (TC) treated for optimum attachment and growth of anchorage-dependent cells. Corning™ TC-Treated Culture Dishes provide an ideal solution for growing and maintaining cells in vitro.
Falcon™ Tissue Culture Dish with Grid Available on GSA/VA Contract for Federal Government customers only.
Features flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Corning™ Falcon™ Easy-Grip Tissue Culture Dishes are uniquely designed to simplify lifting one dish or a small stack of dishes.
Corning™ Transwell™ Multiple Well Plate with Permeable Polycarbonate Membrane Inserts Available on GSA/VA Contract for Federal Government customers only.
Patented self-centering design provides independent access to both apical and basolateral membranes of the cell monolayer. Corning™ Transwell™Multiple Well Plate with Permeable Polycarbonate Membrane Inserts provide an ideal solution for studying anchorage-dependent and -independent cells.
Corning™ Sterile Cell Strainers Available on GSA/VA Contract for Federal Government customers only.
Isolates primary cells rapidly to consistently obtain a uniform single-cell suspension from tissues. Corning™ Sterile Cell Strainers are ideal for preparation of flow cytometry samples and are used with stem cells and primary cells.
MP Biomedicals™ Roll & Grow™ Plating Beads
Safely and easily spread yeast and bacterial cells
MilliporeSigma™ Millicell™ Culture Plate Inserts Available on GSA/VA Contract for Federal Government customers only.
Designed for improved cell differentiation and expanded applications involving analytical cell biology. Millicell™ Culture Plate Inserts incorporate PTFE Biopore™ membranes with 0.4μm pores. Ready-to-use 30 or 12mm inserts for 6 and 24-well plates.
Bel-Art™ Cloning Cylinders Available on GSA/VA Contract for Federal Government customers only.
Manufactured from polystyrene. SP Scienceware™ Cloning Cylinders facilitates the isolation of individual cell clones.
Bel-Art™ ProCulture Cell Lifter Available on GSA/VA Contract for Federal Government customers only.
Economical and reusable tools to scrape cell lysates or remove and harvest tissue cultures from all types of dishes
Falcon™ Cell Culture Inserts: For Use With 6-Well Plates Available on GSA/VA Contract for Federal Government customers only.
Cell culture on permeable supports results in greater morphological and/or physiological differentiation of many cell types
Bel-Art™ Cloning Discs
Save time and reduce effort when isolating a large number of clones
Corning™ BioCoat™ Matrigel™ Invasion Chamber: With GFR Matrigel Matrix Available on GSA/VA Contract for Federal Government customers only.
Provide rapid and reproducible quantification of cell invasion in vitro
Thermo Scientific™ Nunc™ Cell Scrapers Available on GSA/VA Contract for Federal Government customers only.
Conduct a variety of applications with sterile, nonpyrogenic Thermo Scientific™ Nunc™ Cell Scrapers, which feature an adjustable blade.
Corning™ Ultra-Low Attachment Dishes Available on GSA/VA Contract for Federal Government customers only.
Unique covalently bonded hydrogel surface is hydrophilic and neutrally charged
Bel-Art™ SP Scienceware™ Flowmi™ Cell Strainers for 1000 μL Pipette Tips Available on GSA/VA Contract for Federal Government customers only.
Provides fast and efficient filtering of small volume samples (up to 1000 μL) prior to FLOW or FACS analysis (patent pending). Bel Art™ SP Scienceware™ Flowmi™ Cell Strainers for 1000 μL Pipette Tips are sterile and come in two porosity sizes.
Thermo Scientific™ BioLite Cell Culture Treated Dishes Available on GSA/VA Contract for Federal Government customers only.
Perform general cell culture procedures more economically with Thermo Scientific™ BioLite Cell Culture Treated Dishes, guaranteed to be sterile, non-pyrogenic and non-cytotoxic.
Corning™ Snapwell™ Culture Inserts Available on GSA/VA Contract for Federal Government customers only.
Designed with a ring supported membrane for use with diffusion or Ussing chambers once cells are grown to confluence. Corning™ Snapwell™ Culture Inserts are primarily used for transport and electrophysiological studies.
  spinner