Ammonium Hydroxide
Ammonium Hydroxide (Certified ACS Plus), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonium Hydroxide Name Note: about 14.8N CAS: 7664-41-7 CAS: 7732-18-5 Purity Grade: Certified ACS Plus Grade Assay: 28.0 to 30.0% (w/w) Assay Percent Range: Assay: 28.0 to 30.0 w/w % Molecular Formula: H5NO Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00011418 Synonym: Aqueous Ammonia
Ammonium hydroxide, for analysis, 20% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonium hydroxide CAS: 1336-21-6 CAS: 7732-18-5 Purity Grade: For analysis Assay: ≥20% (Acidimetry) Molecular Formula: H5NO Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066508
Ammonium Hydroxide (Optima™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonium Hydroxide CAS: 1336-21-6 Purity Grade: OPTIMA™ Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066650 Synonym: Aqueous Ammonia
Ammonia, ca. 7N solution in methanol, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonia Name Note: ca. 7N Solution in Methanol CAS: 7664-41-7 CAS: 67-56-1 Molecular Formula: H3N Linear Formula: NH3 Formula Weight: 17.03 MDL Number: MFCD00011418
Ammonium Hydroxide, ACS, 28.0-30.0%NH™, Alfa Aesar
Chemical Name or Material: Ammonium hydroxide CAS: 1336-21-6 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: 28.0-30.0% NH3 Molecular Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066650
Ammonium hydroxide, 25% NH3, 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Ammonium hydroxide CAS: 1336-21-6 Physical Form: Liquid Assay Percent Range: 99.99% (metals basis) Molecular Formula: H5NO Formula Weight: 35.05 MDL Number: MFCD00066650
Ammonia, 2M solution in methanol, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonia CAS: 7664-41-7 CAS: 67-56-1 Molecular Formula: H3N Linear Formula: NH3 Formula Weight: 17.03 MDL Number: MFCD00011418
Ammonium hydroxide, extra pure, 25% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonium hydroxide CAS: 1336-21-6 CAS: 7732-18-5 Purity Grade: Extra Pure Assay: ≥25% (as NH3) (total base) Molecular Formula: H5NO Linear Formula: NH4OH Formula Weight: 35.05
Ammonium Hydroxide (26° Bé/NF/FCC), Fisher Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 7664-41-7 CAS: 7732-18-5 Purity Grade: FCC/NF Assay: 27.0 to 31.0% (NF) Molecular Formula: H5NO Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066650 Synonym: Aqueous Ammonia
Ammonium Hydroxide (TraceMetal™ Grade), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonium Hydroxide CAS: 1336-21-6 Purity Grade: TraceMetal Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066650 Synonym: Aqueous Ammonia
Ammonium hydroxide, ACS reagent, 28-30% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ammonium hydroxide CAS: 1336-21-6 CAS: 7732-18-5 Purity Grade: ACS Reagent Grade Assay: 28.0 to 30.0% (HCl-titration (as NH3)) Molecular Formula: H5NO Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066650
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 28-30 wt% solution of NH3 in water CAS: 1336-21-6 CAS: 7732-18-5 Purity Grade: For analysis Molecular Formula: H5NO Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066650
Ammonium hydroxide, for analysis, 25% solution in water, ACROS Organics™
Chemical Name or Material: Ammonium hydroxide CAS: 1336-21-6 CAS: 7732-18-5 Purity Grade: For analysis Molecular Formula: H5NO Linear Formula: NH4OH Formula Weight: 35.05 MDL Number: MFCD00066650
Ammonium Hydroxide, 1.00 Normal, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 7732-18-5 CAS Min %: 92.0 CAS Max %: 96.0 CAS: 1336-21-6 CAS Min %: 5.73 CAS Max %: 5.96 Physical Form: Liquid
Ammonium Hydroxide, 25% (v/v) Aqueous Solution (1+3), Ricca Chemical
CAS: 7732-18-5 Physical Form: Liquid Assay Percent Range: 25% (v/v) Aqueous Solution (1 + 3)
Ammonium Hydroxide, 10% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 7732-18-5 CAS Min %: 90.0 CAS Max %: 94.0 CAS: 1336-21-6 CAS Min %: 9.0 CAS Max %: 11.0 Physical Form: Liquid
Ammonium Hydroxide, 50% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 7732-18-5 CAS Min %: 49.0 CAS Max %: 51.0 CAS: 1336-21-6 CAS Min %: 49.0 CAS Max %: 51.0 Physical Form: Liquid
Ammonium hydroxide, Macron Fine Chemicals™
Chemical Name or Material: Ammonium hydroxide CAS: 1336-21-6 Physical Form: Liquid Molecular Formula: NH4OH Formula Weight: 35.04 Synonym: Ammonia water
Ammonium Hydroxide, 28.0% to 30.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Ammonium Hydroxide CAS: 1336-21-6 Purity Grade: Baker Analyzed™ ACS Reagent Molecular Formula: NH4OH Formula Weight: 35.05
Ammonia Solution, Strong; Ammonium Hydroxide, N.F., A.C.S., J.T.Baker™
Chemical Name or Material: Ammonia Solution Name Note: Strong; Ammonium Hydroxide Purity Grade: ACS/NF Molecular Formula: NH3 Formula Weight: 17.03
Ammonium Hydroxide, 20% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 1336-21-6 (Ammonium Hydroxide) CAS: 7732-18-5 (Water, Deionized) Physical Form: Liquid
Ammonium Hydroxide, 28.0 to 30.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Ammonium Hydroxide CAS: 1336-21-6 Purity Grade: Baker Analyzed™ ACS Reagent Molecular Formula: NH4OH Formula Weight: 35.05
Ammonium Hydroxide, 0.2, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™
Chemical Name or Material: Ammonium Hydroxide CAS: 1336-21-6 Purity Grade: BAKER INSTRA-ANALYZED™ Plus Molecular Formula: NH4OH Formula Weight: 35.05
Ammonia Dilute R1, Ricca Chemical
Chemical Name or Material: Ammonia Dilute R1, Ricca Chemical CAS: 7732-18-5 CAS Min %: 72.0 CAS Max %: 75.0 CAS: 1336-21-6 CAS Min %: 26.21 CAS Max %: 27.28 Purity Grade: EP Reagent 1004702
Ammonium Hydroxide, 6.00N, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide, 6.00N, Ricca Chemical CAS: 7732-18-5 CAS Min %: 71.0 CAS Max %: 74.0 CAS: 1336-21-6 CAS Min %: 26.6 CAS Max %: 27.69
Ammonium Hydroxide, 30%, BAKER INSTRA-ANALYZED Reagent, J.T.Baker™
Chemical Name or Material: Ammonium Hydroxide, 30% CAS: 1336-21-6 Assay Percent Range: 28.0 to 30.0 % Molecular Formula: NH4OH Formula Weight: 35.05
Ammonium Hydroxide, 2.00 Normal, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 7732-18-5 CAS Min %: 87.0 CAS Max %: 91.0 CAS: 1336-21-6 CAS Min %: 10.83 CAS Max %: 11.27 Physical Form: Liquid
Ammonium Hydroxide, 10% (v/v), with HNO3, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 7732-18-5 CAS Min %: 90.0 CAS Max %: 93.0 CAS: 1336-21-6 CAS Min %: 8.08 CAS Max %: 8.41 CAS: 7697-37-2 CAS Min %: 0.13 CAS Max %: 0.14 Physical Form: Acetate
Ammonium Hydroxide, 6.00 Normal, Ricca Chemical
Chemical Name or Material: Ammonium Hydroxide CAS: 7732-18-5 CAS Min %: 71.0 CAS Max %: 74.0 CAS: 1336-21-6 CAS Min %: 26.6 CAS Max %: 27.69 Physical Form: Liquid
  spinner