Methylene Chloride (CH2Cl2)
Methylene Chloride (Stabilized/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride Name Note: Stabilized CAS: 75-09-2 Purity Grade: Certified ACS Reagent Grade Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Dichloromethane, DCM
Methylene Chloride, Optima™, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride CAS: 75-09-2 Purity Grade: OPTIMA™ Purity Grade Notes: High purity mobile phase for HPLC and/or extraction solvent for GC applications Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Dichloromethane, DCM
Methylene Chloride (Pesticide), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride CAS: 75-09-2 Purity Grade: Pesticide Grade Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Dichloromethane, DCM Acidity: Pass Test (USP) Additional Information: Vapor Pressure: Negligible Conductivity: 20 μS/cm max. (EP/BP)
Methylene Chloride (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride Name Note: Stabilized CAS: 75-09-2 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Physical Form: Liquid Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Dichloromethane, DCM
Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane Name Note: Stabilized with Amylene CAS: 75-09-2 CAS: 513-35-9 Purity Grade: For peptide synthesis Assay: 99.9% Assay Percent Range: 99.9% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Methylene Chloride (GC Resolv™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride CAS: 75-09-2 Purity Grade: GC Resolv Purity Grade Notes: High purity solvent for sample prep prior to trace analysis by GC and/or GC/MS Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Dichloromethane, DCM
Dichloromethane, 99+%, extra pure, stabilized with ethanol, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane CAS: 75-09-2 CAS: 64-17-5 Purity Grade: Extra Pure Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane Name Note: Stabilized with Amylene CAS: 75-09-2 CAS: 513-35-9 Purity Grade: ACS Reagent Grade Assay: 99.6% Assay Percent Range: 99.6% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Methylene Chloride (Not Stabilized/HPLC), Fisher Chemical
Chemical Name or Material: Methylene Chloride Name Note: Not Stabilized CAS: 75-09-2 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Physical Form: Liquid Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Dichloromethane, DCM
Methylene Chloride (Stabilized/Spectranalyzed™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride Name Note: Stabilized CAS: 75-09-2 Purity Grade: Spectranalyzed™ Purity Grade Notes: Also meets ACS specifications Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Dichloromethane, DCM
Alfa Aesar™ Dichloromethane, Spectrophotometric Grade, 99.7+%, stab. with amylene
Chemical Name or Material: Dichloromethane CAS: 75-09-2 Name Note: Stabilized with amylene Purity Grade: Spectrophotometric grade Formula Weight: 84.93 MDL Number: MFCD00000881 UN Number: UN1593 Beilstein: 1730800 Merck Index: 14,6063
Dichloromethane, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane Name Note: 99.9% CAS: 75-09-2 CAS: 513-35-9 Purity Grade: For HPLC Assay: ≥99.9% Molecular Formula: C H2 Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, 99.9%, Extra Dry, anhydrous, SC, AcroSeal™, ACROS Organics™
Chemical Name or Material: Dichloromethane Name Note: Anhydrous, SC CAS: 75-09-2 CAS: 513-35-9 Purity Grade: Extra Dry Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene Chloride
Dichloromethane, 99.9%, Extra Dry, AcroSeal™, 99.9%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane Name Note: Anhydrous CAS: 75-09-2 CAS: 513-35-9 Purity Grade: Extra Dry Assay: ≥99.0% Assay Percent Range: 99.9% Molecular Formula: C H2 Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Methylene Chloride (With Cyclohexene Preservative/HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride Name Note: With cyclohexene preservative CAS: 75-09-2 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Dichloromethane, DCM
Dichloromethane, 99.9%, for residue analysis, for anal.of polyarom.hydrocarb., stab.with amylene, ACROS Organics™
Chemical Name or Material: Dichloromethane CAS: 75-09-2 CAS: 513-35-9 Purity Grade: For residue analysis Assay: 99.9% Assay Percent Range: 99.9% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 Synonym: Methylene chloride
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane CAS: 75-09-2 CAS: 513-35-9 Purity Grade: Extra Dry Purity Grade Notes: Water <30ppm Assay: 99.9% Assay Percent Range: 99.9% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, for residue and pestic. anal., stab. with amylene, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane Name Note: ECD Tested, For Pesticide analysis CAS: 75-09-2 CAS: 513-35-9 Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Alfa Aesar™ Dichloromethane, 99+%, stab. with ca. 50ppm amylene
Chemical Name or Material: Dichloromethane CAS: 75-09-2 Name Note: stabilized with ca. 50ppm amylene Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 UN Number: UN1593 Beilstein: 1730800 Merck Index: 14,6063 Synonym: DCM; Methylene chloride
Dichloromethane, Anhydrous, 99.8% min., DriSolv, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane, Anhydrous Name Note: DriSolv™ CAS: 75-09-2 Physical Form: Liquid Assay Percent Range: 99.8%, min. Molecular Formula: CH2Cl2 Linear Formula: K2HPO4 Formula Weight: 174.18 Synonym: Potassium phosphate dibasic
Dichloromethane ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Dichloromethane CAS: 75-09-2 Purity Grade: ACS Reagent Grade Molecular Formula: CH2Cl2 Formula Weight: 84.93
Dichloromethane 99+%, stabilized with amylene, pure, ACROS Organics™
Chemical Name or Material: Dichloromethane Assay Percent Range: 99+% Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, 99.8%, for HPLC, stabilized with methanol, ACROS Organics™
Chemical Name or Material: Dichloromethane Name Note: Stabilized CAS: 75-09-2 CAS: 67-56-1 Purity Grade: HPLC Grade Assay: 99.8% Assay Percent Range: 99.80% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains cyclohexene , >99.9%, Honeywell Burdick & Jackson
Density: 1.33 g/cm3 Boiling Point: 40°C Formula Weight: 84.94 Purity Grade: for HPLC CAS: 75-09-2 Chemical Name or Material: Dichloromethane CAS Max %: 100.0000% Beilstein: 1730800 Name Note: B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains cyclohexene , >99.9% Packaging: Glass Bottle Assay: >99.9% UN Number: UN1593
Dichloromethane, 99.8+%, for analysis, stabilized with amylene, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane CAS: 75-09-2 CAS: 513-35-9 Purity Grade: For analysis Assay: ≥99.8;% Assay Percent Range: 99.8+% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, For pesticide residue analysis, Honeywell Riedel-de Haën™
Beilstein: 1730800 CAS: 75-09-2 Chemical Name or Material: Dichloromethane Density: 1.325 g/mL at 25 °C(lit.) Assay: >99.8% Formula Weight: 84.93 Purity Grade: for pesticide residue analysis MDL Number: MFCD00000881 UN Number: UN1593
Dichloromethane ChromAR, Macron Fine Chemicals™
Chemical Name or Material: Dichloromethane CAS: 75-09-2 Molecular Formula: CH2Cl2 Formula Weight: 84.93
Dichloromethane, 99.8%, for HPLC, stabilized with amylene, ACROS Organics™
Chemical Name or Material: Dichloromethane Name Note: Stabilized CAS: 75-09-2 CAS: 513-35-9 Purity Grade: For HPLC Assay: 99.8% Assay Percent Range: 99.8% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, Stabilized, OmniSolv™, 99.9%, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane, Stabilized, 99.9% CAS: 75-09-2 Formula Weight: 84.93
  spinner