Desiccants and Drying Agents
Silica Gel Desiccant (100-200 Mesh/923, Chromatographic Grade), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.083 MDL Number: MFCD00011232 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Drierite, with indicator, 8 mesh, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.134 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium;sulfate SMILES: [O-]S(=O)(=O)[O-].[Ca+2]
Silica Gel Desiccant (28-200 mesh, Grade 12/Certified ACS), Fisher Chemical
CAS: 63231-67-4 Molecular Formula: H2O3Si Molecular Weight (g/mol): 78.098 MDL Number: MFCD00011232 InChI Key: IJKVHSBPTUYDLN-UHFFFAOYSA-N PubChem CID: 14768 IUPAC Name: dihydroxy(oxo)silane SMILES: O[Si](=O)O
W.A. Hammond Drierite™ Indicating Absorbents
Efficient desiccant giving constant visual indication of the progress of absorption and exhaustion. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.
Drierite™, with indicator, 10-20 mesh, ACROS Organics™
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.134 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium;sulfate SMILES: [O-]S(=O)(=O)[O-].[Ca+2]
Silica Gel Desiccant (6-12 Mesh, Certified ACS), Fisher Chemical
CAS: 63231-67-4 Molecular Formula: H2O3Si Molecular Weight (g/mol): 78.098 MDL Number: MFCD00011232 InChI Key: IJKVHSBPTUYDLN-UHFFFAOYSA-N PubChem CID: 14768 IUPAC Name: dihydroxy(oxo)silane SMILES: O[Si](=O)O
Calcium Chloride, Anhydrous (Pellets, 4-20 mesh, for Desiccators), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
CAS: 10043-52-4 Molecular Formula: CaCl2 Molecular Weight (g/mol): 110.978 MDL Number: MFCD00010903 InChI Key: UXVMQQNJUSDDNG-UHFFFAOYSA-L PubChem CID: 5284359 ChEBI: CHEBI:3312 IUPAC Name: calcium;dichloride SMILES: [Cl-].[Cl-].[Ca+2]
Silica gel, for column chrom., ultrapure, 60-200μ, 60A, ACROS Organics™
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.083 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N Synonym: Silicon dioxide PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Alfa Aesar™ Silica gel 60, 0.060-0.2mm (70-230 mesh)
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.083 MDL Number: MFCD00011232 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Silica Gel Desiccant, Indicating, 6 to 18 Mesh, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
CAS: 1343-98-2 Molecular Formula: H4O4Si Molecular Weight (g/mol): 96.113 InChI Key: RMAQACBXLXPBSY-UHFFFAOYSA-N PubChem CID: 14942 ChEBI: CHEBI:26675 IUPAC Name: silicic acid SMILES: O[Si](O)(O)O
W.A. Hammond Drierite™ Drying Desiccants
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.134 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L Synonym: Abosrbent PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium;sulfate SMILES: [O-]S(=O)(=O)[O-].[Ca+2]
DRIERITE™, Indicating, 4 Mesh, J.T.Baker™
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.134 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium;sulfate SMILES: [O-]S(=O)(=O)[O-].[Ca+2]
Silicagel orange, for drying purposes, non toxic grade, ACROS Organics™
CAS: 1327-36-2 Molecular Formula: Al2O5Si Molecular Weight (g/mol): 162.04 MDL Number: MFCD00077645 InChI Key: HNPSIPDUKPIQMN-UHFFFAOYSA-N
Drierite™, without indicator, 8 mesh, ACROS Organics™
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.134 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium;sulfate SMILES: [O-]S(=O)(=O)[O-].[Ca+2]
Silica Gel Desiccant (60-200 Mesh/Chromatographic Grade), Fisher Chemical
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.083 MDL Number: MFCD00011232 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Florisil, 60-100 mesh, for column chromatography, ACROS Organics™
CAS: 1343-88-0 Molecular Formula: MgO3Si Molecular Weight (g/mol): 100.39 MDL Number: MFCD00078249 InChI Key: ZADYMNAVLSWLEQ-UHFFFAOYSA-N
Silica Gel Indicating Desiccants, -6+16 mesh granules, Alfa Aesar™
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.083 MDL Number: MFCD00011232 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Drierite™, with indicator, 4 mesh, ACROS Organics™
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.134 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium;sulfate SMILES: [O-]S(=O)(=O)[O-].[Ca+2]
W.A. Hammond Drierite™ Drying Tubes
Chemical Name or Material: Drierite Indicating Drying Tube, WA Hammond
Desiccant, Silica Gel/Cobaltous Chloride, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Desiccant, Silica Gel/Cobaltous Chloride
Desiccant, EMD Chemicals Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Desiccant Name Note: 8% Indicating, 8 Mesh Molecular Formula: SiO2
Soda Lime, Indicating Type, 4 to 8 Mesh, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
CAS: 8006-28-8 Molecular Formula: CaHNaO2 Molecular Weight (g/mol): 96.074 InChI Key: HUAUNKAZQWMVFY-UHFFFAOYSA-M PubChem CID: 66545795 IUPAC Name: sodium;oxocalcium;hydroxide SMILES: [OH-].O=[Ca].[Na+]
Calcium Chloride (Desiccant, 20 mesh and Finer/Certified) ACS, Fisher Chemical
CAS: 10043-52-4 Molecular Formula: CaCl2 Molecular Weight (g/mol): 110.978 MDL Number: MFCD00010903 InChI Key: UXVMQQNJUSDDNG-UHFFFAOYSA-L PubChem CID: 5284359 ChEBI: CHEBI:3312 IUPAC Name: calcium;dichloride SMILES: [Cl-].[Cl-].[Ca+2]
Soda Lime, ACS, Alfa Aesar
CAS: 8006-28-8 Molecular Formula: CaHNaO2 Molecular Weight (g/mol): 96.074 InChI Key: HUAUNKAZQWMVFY-UHFFFAOYSA-M PubChem CID: 66545795 IUPAC Name: sodium;oxocalcium;hydroxide SMILES: [OH-].O=[Ca].[Na+]
  spinner