Inoculating Loops and Needles
Thermo Scientific™ Nunc™ Disposable Loops and Needles Available on GSA/VA Contract for Federal Government customers only.
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Fisherbrand™ Disposable Inoculating Loops and Needles, Flexible Needle and Loop Edge
For quantitative procedures such as sampling, serial dilutions and urine counts, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 9 in. (23cm) are molded from a specially formulated, biologically inert polymer.
BD Difco™ Disposable Inoculating Needles and Loops Edge Available on GSA/VA Contract for Federal Government customers only.
Sterile, disposable inoculating needles do not require flaming
Fisherbrand™ Disposable Inoculating Loops and Needles
Designed for quantitative procedures such as sampling, serial dilutions, as well as for bacterial inoculation. Fisherbrand™ Disposable Inoculating Loops and Needles, Length: 8 in. (20cm) are precision molded from a specially formulated, biologically inert polymer.
Copan Diagnostics Calibrated Disposable Inoculating Loops and Needles in Zip Lock Pack Edge Available on GSA/VA Contract for Federal Government customers only.
Calibrated 1 and 10μL plastic disposable inoculating loops and needles
Fisherbrand™ Nichrome/Aluminum Inoculating Loops Edge
Fisherbrand Nichrome/Aluminum Inoculating Loops consist of a nichrome wire loop mounted to an aluminum handle. Inoculating loops are used to introduce microorganisms into a culture medium.
Globe Scientific Sterile Rigid Inoculating Loops with Needles
These disposable inoculation loops and needles offer the most convenient and safest way of streaking culture media. Molded from a special medical grade of polystyrene (PS), they offer a strong, rigid feel to provide comfortable, stable and uniform streaking.
Decon™ LeStreak™ Microstreakers Available on GSA/VA Contract for Federal Government customers only.
Perfect for inoculating or transferring microbiological samples
Thermo Scientific™ Nunc™ Disposable Loops and Needles, needle Available on GSA/VA Contract for Federal Government customers only.
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Troemner™ Inoculating Turntable Available on GSA/VA Contract for Federal Government customers only.
Hand-operated inoculating turntables
Pro-Lab Diagnostics™ Pro-Loops™ Insulated Inoculating Loop Wire Holders
Available in nickel-plated brass or lightweight aluminum
Decon™ LeLoop™ Inoculating Loops Available on GSA/VA Contract for Federal Government customers only.
More economical than platinum or disposable plastic loops
Globe Scientific Sterile Rigid 10uL & 1uL Combo Inoculation Loops
These disposable inoculation loops and needles offer the most convenient and safest way of streaking culture media. Molded from a special medical grade of polystyrene (PS), they offer a strong, rigid feel to provide comfortable, stable and uniform streaking.
Decon™ LeLoop™ Holder Available on GSA/VA Contract for Federal Government customers only.
Holders for LeLoop Inoculating Loops
Bio Plas™ Astral™ Inoculation Device System
Economical, disposable inoculation system designed for safety and dependable sterility
Thermo Scientific™ Nunc™ Disposable Loops and Needles, needle Available on GSA/VA Contract for Federal Government customers only.
Easily identify these color-coded disposable loops and needles, precision molded from nontoxic (USP Class VI) polystyrene material.
Leica Microsystems Covidien No. 26 Platinum-iridium Loops
Loop, Platinum-iridium, Covidien; Fused, 3mm I.D., 2-3/4in. long, Without holder
Thermo Scientific™ Reusable Calibrated Inoculating Loops Available on GSA/VA Contract for Federal Government customers only.
Select, inoculate or transfer a well-defined microbiological sample volume on media plates with these reusable calibrated inoculating loops.
Corning™ Sterile Polystyrene Inoculating Loops Available on GSA/VA Contract for Federal Government customers only.
Constructed of flexible polystyrene for optimum retrieval of an inoculum from a culture. Gosselin™ Sterile Polystyrene Inoculating Loops provide a superior solution for transferring microscopic organisms, inoculum, or streaking.
Decon™ LeStab™ Inoculating Microneedles Available on GSA/VA Contract for Federal Government customers only.
Perfectly balanced for easy manipulation
Thermo Scientific™ Remel™ Calibrated Inoculating Loops Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Calibrated Inoculating Loops come in a variety of materials: platinum, twisted nichrome and plastic.Some with color coded with insulated handle.
Bel-Art™ SP Scienceware™ Replica-Plating Tool and Replacement Velveteen Squares Available on GSA/VA Contract for Federal Government customers only.
Utilize a locking ring which secures sterile velveteen squares onto a PVC cylinder for easy control of the plating device
Bel-Art™ SP Scienceware™ Replacement Cushion for Inoculating Turntable
Replacement pad for a lightweight and portable inoculating turntable.
Leica Microsystems Inoculating Loops and Holders
Loops and holders designed for culturing, transferring, streaking or isolating tissue cultures in cytology or histology
Thermo Scientific™ Remel™ Microloop
Thermo Scientific™ Remel Microloop is a twisted nichrome loop without handle.
  spinner