Personal Protective Equipment
Filtered Search Results
Fisherbrand™ Comfort Nitrile Gloves with Aloe and Chamomile
Soft nitrile protection with aloe and chamomile.
| Disposable | Single-use |
|---|---|
| Length (Metric) | 250 mm |
| Applications | Healthcare,Industrial,Laboratory |
| Chemotherapeutic Rated | Yes |
| Color | Blue,Green |
| Length (English) | 9.8 in. |
| Quantity | 100 Gloves/Box |
| Material | Nitrile |
| Powdered | Powder Free |
| Sterility | Non-sterile |
| Finger Thickness (Metric) | 0.12 mm |
| Thickness (Metric) | Palm: 0.09 mm, Finger: 0.12 mm |
| FDA 510 (k) | Yes |
| Product Type | Nitrile Examination Glove |
| Latex Free | Yes |
| Exterior Finish | Fingertip Textured |
| Certifications/Compliance | ASTM D6319,ASTM D6978,FDA21 CFR 177-2600, US Food Contact Approved,US QSR/FDA 510(k) Medical Examination Grade |
| AQL | 1.5 |
| Glove Cuff Style | Beaded |
| Palm Thickness (Metric) | 0.09 mm |
Fisherbrand™ Spunbond Polypropylene Non-Skid Shoe Covers
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Shoe covers feature non-skid soles for greater traction.
| Disposable | Single-use |
|---|---|
| Sole Type | Non-skid |
| Product Type | Shoe Covers |
| Color | Blue |
| Latex Free | Yes |
| Material | Spunbond Polypropylene |
| Closure Type | Elastic Ankle |
| Sterility | Non-sterile |
Ansell Kimtech™ Purple Nitrile™ Gloves
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Purple nitrile glove offers maximum protection for higher-risk procedures. Unique color differentiates from latex and other synthetic gloves.
| Tensile Strength | 21 MPa |
|---|---|
| Disposable | Single-use |
| Length (Metric) | 24.13 cm |
| Applications | Laboratory,Healthcare |
| Chemotherapeutic Rated | Yes |
| Color | Purple |
| Length (English) | 9.5 in. |
| Material | Nitrile |
| Powdered | Powder Free |
| Finger Thickness (Metric) | 0.15 mm |
| Thickness (Metric) | 0.15 mm |
| Series | Kimtech™ |
| FDA 510 (k) | Yes |
| Latex Free | Yes |
| Exterior Finish | Partially Textured |
| Certifications/Compliance | ASTM D6978-05 (non-sterile only), Meets USP <800> guidelines |
| AQL | 0.65 |
| Palm Thickness (Metric) | 0.12 mm |
| Lens Tint | Clear |
|---|---|
| Shape | Rectangle |
| Lens Coating | Hardcoat |
| Product Type | Safety Glasses |
| Certifications/Compliance | ANSI Z87.1+, ANSI Z80.3 |
| Temple Style | Adjustable; spatula |
Fisherbrand™ Powder Free Dark Blue Nitrile Gloves
Choose a latex-free glove with exceptional durability and puncture resistance. Fisherbrand™ powder free Nitrile Exam Gloves provide a superior fit, comfort, tactile sensitivity and dexterity.
| Disposable | Single-use |
|---|---|
| Length (Metric) | 240 mm |
| Applications | Healthcare,Industrial,Laboratory |
| Chemotherapeutic Rated | No |
| Color | Dark Blue |
| Length (English) | 9.5 in. |
| Quantity | 100 Gloves/Box |
| Material | Nitrile |
| Powdered | Powder Free |
| Sterility | Non-sterile |
| Finger Thickness (Metric) | 0.14 mm |
| Thickness (Metric) | Palm: 0.12 mm, Finger: 0.14 mm |
| FDA 510 (k) | No |
| Latex Free | Yes |
| Exterior Finish | Fully Textured |
| Certifications/Compliance | ASTM D6319,FDA21 CFR 177-2600-US Food Contact Approved |
| AQL | 1.5 |
| Palm Thickness (Metric) | 0.12 mm |
Fisherbrand™ Unisex Cotton Lab Coats
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Open cuffs provide comfortable and unrestrictive fit.
| Disposable | Reusable |
|---|---|
| Cuff Style | Open |
| Product Type | General Purpose Lab Coat |
| Color | White |
| Material | Cotton |
| Gender | Unisex |
| Closure Type | Snaps |
| Collar Type | Standard Pointed |
| Sterility | Non-sterile |
| No. of Pockets | 3 |
Fisherbrand™ Visitorspec Safety Glasses, Coated, Antifog
Ideal for protecting visitors in industrial or medical settings.
Fisherbrand™ Disposable Lab Coats
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
White 50 gsm disposable polypropylene lab coats that provide breathable protection with a turndown collar, elastic cuff and hook-and-loop closure.
| Thickness | 50 gsm |
|---|---|
| Disposable | Single-use |
| Cuff Style | Elastic |
| Product Type | General Purpose Lab Coat |
| Packaging Type | 5 packs of 10 |
| Color | White |
| Material | Polypropylene |
| Gender | Unisex |
| Closure Type | Hook and Loop |
| Collar Type | Turn Down |
| Sterility | Non-sterile |
Fisherbrand™ Disposable SMS Laboratory Jackets
The Fisherbrand™ Disposable SMS Lab Jackets with a knitted collar and cuffs. Designed to protect healthcare professionals, lab technicians and other users from fluids and cross-contamination.
| Disposable | Single-use |
|---|---|
| Packaging Type | Bagged |
| Material | SMS |
| Gender | Unisex |
| Sterility | Non-sterile |
| No. of Pockets | 3 |
| Resistance | Liquids |
| Cuff Style | Knit |
| Product Type | General Purpose Lab Coat |
| Latex Free | Yes |
| Style | Hip-length |
| No. per Pack | 10/Pk. |
| Certifications/Compliance | ANSI, AAMI PB70 |
| Autoclavable | Not Autoclavable |
| Closure Type | Snaps |
| Collar Type | Knit |
Fisherbrand™ Comfort Nitrile Gloves
Choose a glove with enhanced comfort and tactile sensitivity. Fisherbrand™ Comfort Nitrile Gloves are powder free and latex-free with a beaded fit and textured fingers.
| Disposable | Single-use |
|---|---|
| Applications | Healthcare,Industrial,Laboratory |
| Chemotherapeutic Rated | Yes |
| Color | Blue |
| Length (English) | 9.8 in. |
| Material | Nitrile |
| Powdered | Powder Free |
| Sterility | Non-sterile |
| Hand | Ambidextrous |
| Finger Thickness (Metric) | 0.09 mm |
| Thickness (Metric) | Palm: 0.07 mm, Finger: 0.09 mm |
| FDA 510 (k) | Yes |
| Latex Free | Yes |
| Exterior Finish | Fingertip Textured |
| Certifications/Compliance | ASTM D6319,ASTM D6978,ISO 10993-10:2002, ISO 9001, ISO 13485,FDA21 CFR 177- 2600-US Food Contact Approved,US QSR/FDA 510(k) Medical Examination Grade |
| AQL | 1.5 |
| Palm Thickness (Metric) | 0.07 mm |
Fisherbrand™ Controlled Environment Coverall
Fisherbrand microporous composite coveralls are designed for many non-hazardous applications and will keep the wearer clean and comfortable all day.
| Disposable | Single-use |
|---|---|
| Neck Style | Collared |
| Color | White |
| Quantity | 25/Cs. |
| Sleeve Style | Set-in Sleeve |
| Waist Style | Standard |
| Material | Microporous Composite Fabric |
| Gender | Unisex |
| For Use With (Application) | Controlled and Critical Environments |
| Sterility | Non-sterile |
| No. of Pockets | None |
| Resistance | Particle |
| Seam Type | Serged |
| Cuff Style | Elastic |
| Product Type | Cleanroom Coveralls |
| Cleanroom Class | Class 100 (ISO 5) |
| Back Style | Elastic Back |
| Latex Free | Yes |
| Ankle Style | Elastic |
| Hood Type | Unhooded |
| Closure Type | Zipper |
| Collar Type | Tapered |
Fisherbrand™ Extended Cuff Nitrile Gloves
Choose a glove with exceptional durability and added protection. Fisherbrand™ extended cuff nitrile gloves are extra-thick with textured fingertips for a superior grip and are tested for use with chemotherapy drug exposure.
| Disposable | Single-use |
|---|---|
| Length (Metric) | 300 mm |
| Applications | Healthcare,Industrial,Laboratory |
| Chemotherapeutic Rated | Yes |
| Color | Teal |
| Length (English) | 11.8 in. |
| Quantity | 50 Gloves/Box |
| Material | Nitrile |
| Powdered | Powder Free |
| Sterility | Non-sterile |
| Hand | Ambidextrous |
| Finger Thickness (Metric) | 0.19 mm |
| Thickness (Metric) | Palm: 0.15 mm, Finger: 0.19 mm |
| FDA 510 (k) | Yes |
| Latex Free | Yes |
| Exterior Finish | Fingertip Textured |
| Certifications/Compliance | ASTM D6319,ASTM D6978,FDA21 CFR 177-2600-US Food Contact Approved,US QSR/FDA 510(k) Medical Examination Grade |
| AQL | 1.5 |
| Palm Thickness (Metric) | 0.15 mm |
Fisherbrand™ String Knit Gloves with PVC Dot Grip
PVC dots enhance grip capabilities and abrasion resistance.
| Disposable | Reusable |
|---|---|
| Grip Design | Dots |
| Construction | String Knit |
| Length (Metric) | 22.6 cm |
| Applications | Industrial |
| Color | Natural,Black |
| Length (English) | 8.9 in. |
| Cut Resistance Rating | ANSI A1 |
| Material | Cotton |
| Powdered | Powder Free |
| Sterility | Non-sterile |
| Hand | Specific |
| Product Type | String Knit Gloves |
| Exterior Finish | Grip Pattern |
| Glove Cuff Style | Continuous Knit |