Antibiotics
Gibco™ Hygromycin B (50mg/mL) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Water-soluble antibiotic purified from bacterium Streptomyces hydroscopicus
Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate) (50mg/mL) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Produced by bacterium Micromonospora rhodorangea and acts by binding ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells
Gibco™ Blasticidin S HCl (10mg/mL), Sterile Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Selective antibiotic in concentration range of 50G–100μg/mL
Gibco™ Sterile Puromycin Dihydrochloride Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Aminonucleoside antibiotic produced by Streptomyces alboniger
Gibco™ Penicillin-Streptomycin (10,000 U/mL) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
For prevention of bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria
Ampicillin Sodium Salt (Crystalline Powder), Fisher BioReagents
Chemical Name or Material: Ampicillin Sodium Salt CAS: 69-52-3 Physical Form: Solid Molecular Formula: C16H18N3O4SNa Formula Weight: 371.39 Synonym: Ampicillin
Gibco™ Molecular Probes™ Gentamicin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea
Gibco™ Antibiotic-Antimycotic (100X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Anti-mycotic, antibiotic for prevention of cell culture contamination
Kanamycin Sulfate (White Powder), Fisher BioReagents
Chemical Name or Material: Kanamycin Sulfate CAS: 25389-94-0 Physical Form: Solid Molecular Formula: C18H36N4O11 · H2SO4 Formula Weight: 582.58
Gibco™ Zeocin™ Zeocin Selection Reagent Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Selection antibiotic, formulation of phleomycin D1, a glycopeptide antibiotic produced by Streptomyces verticillus
Carbenicillin (Disodium Salt), Fisher BioReagents
Chemical Name or Material: Carbenicillin Name Note: Disodium Salt CAS: 4800-94-6 Physical Form: Solid Assay Percent Range: ≥92 % Molecular Formula: C17H16N2Na2O6S Formula Weight: 422.4 Synonym: α-Carboxybenzylpenicillin
Corning™ Penicillin-Streptomycin Solution Available on GSA/VA Contract for Federal Government customers only.
Effective against Gram-positive and Gram-negative bacteria. Corning™ Penicillin-Streptomycin Solution is a mix of the antibiotics Penicillin (10,000 IU) and Streptomycin (10,000µg/mL) in a 100-fold working concentration.
G418 Sulfate (White to Off-white Powder), Fisher BioReagents
Chemical Name or Material: G418 Sulfate CAS: 108321-42-2 Physical Form: Solid Assay Percent Range: > 98 % Molecular Formula: C20H40N4O10 · 2 H2SO4 Formula Weight: 692.7
Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate), Powder Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Produced by bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells
Carbenicillin, Disodium Salt, Powder Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Carbenicillin, Disodium Salt, Powder Name Note: Research Use Only, Not for IVD Use Physical Form: Powder
Sulfanilamide Certified ACS, Fisher Chemical
Chemical Name or Material: Sulfanilamide CAS: 63-74-1 Purity Grade: Certified Grade Physical Form: Solid Assay: ≥98% Assay Percent Range: ≥98% Molecular Formula: C6H8N2O2S Linear Formula: 4-NH2C6H4SO2NH2 Formula Weight: 172.21 MDL Number: MFCD00007939 Synonym: 4-Aminobenzenesulfonamide, P-Anilinesulfonamide
Gibco™ Blasticidin S HCl, powder Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Bacterial selective antibiotic in concentration range of 50G–100μg/mL
Corning™ G418 Sulfate Available on GSA/VA Contract for Federal Government customers only.
Blocks polypeptide synthesis by hindering protein elongation. Corning™ G418 Sulfate is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.
Chloramphenicol (Crystalline Powder), Fisher BioReagents
Chemical Name or Material: Chloramphenicol CAS: 56-75-7 Physical Form: Solid Assay Percent Range: 97% Molecular Formula: C11H12Cl2N2O5 Formula Weight: 323.13 Synonym: Chloromycetin, D-(?)-threo-2, 2-Dichloro-N-[?-hydroxy-?-(hydroxymethyl)-?-(4-nitrophenyl)ethyl]acetamide
Gibco™ Penicillin-Streptomycin-Glutamine (100X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
for prevention from bacterial contamination in cell cultures due to their effective combined action against gram-positive and gram-negative bacteria.
Tetracycline Hydrochloride, Fisher BioReagents
Chemical Name or Material: Tetracycline Hydrochloride CAS: 64-75-5 Physical Form: Solid Molecular Formula: C22 H24N2O8 · HCl Formula Weight: 480.9
Corning™ Hygromycin B Solution Available on GSA/VA Contract for Federal Government customers only.
Blocks polypeptide synthesis and inhibits elongation.
Vancomycin hydrochloride, >900 microgram/mg, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Vancomycin hydrochloride Assay: ≥80.0% (vancomycin B) (HPLC), 9.0% (individual impurities) max., greater than 900μg/mg (potency, on anhydrous substance) Molecular Formula: C66H75Cl2N9O24·HCl Formula Weight: 1485.73
Gibco™ Fungizone™ Fungizone Antimycotic Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Formulation of amphotericin B with sodium deoxycholate as an emulsifying agent to maintain the suspension
Corning™ Gentamicin Sulfate Available on GSA/VA Contract for Federal Government customers only.
Packaged in plastic bottles
Corning™ Antibiotic-Antimycotic Solution Available on GSA/VA Contract for Federal Government customers only.
Packaged in plastic bottles
Amphotericin B (Yellow Solution/250μg/mL), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Amphotericin B Name Note: 250μg/mL CAS: 1397-89-3 Physical Form: Liquid Molecular Formula: C47H73NO17 Formula Weight: 924.1
Corning™ G418 Sulfate, Powder Available on GSA/VA Contract for Federal Government customers only.
Packaged in plastic bottles
Gibco™ Penicillin-Streptomycin (5,000 U/mL) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
For prevention from bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria.
Ionomycin (Calcium Salt), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ionomycin, Calcium Salt Name Note: From Streptomyces conglobatus CAS: 56092-82-1 Assay Percent Range: ≥98.0 % Molecular Formula: C41H70O9Ca Formula Weight: 747.1
  spinner