Ia - Kz Restriction Enzymes
Thermo Scientific™ FastDigest KpnI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest Kpn2I Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ I-SceI Available on GSA/VA Contract for Federal Government customers only.
Cut at TAGGGATAA^CAGGGTAAT sites with I-SceI homing enzyme, which performs best at 37°C in Tango buffer.
Thermo Scientific™ KpnI Available on GSA/VA Contract for Federal Government customers only.
The KpnI restriction enzyme recognizes GGTAC^C sites and cuts best at 37°C in its own unique buffer (Isoschizomers: Asp718I).
Thermo Scientific™ KspAI (HpaI) Available on GSA/VA Contract for Federal Government customers only.
The KspAI (HpaI) restriction enzyme recognizes GTT^AAC sites and cuts best at 37°C in B buffer (Isoschizomers: HpaI).
Thermo Scientific™ Kpn2I (BspEI) Available on GSA/VA Contract for Federal Government customers only.
The Kpn2I (BspEI) restriction enzyme recognizes T^CCGGA sites and cuts best at 55°C in Tango buffer (Isoschizomers: AccIII, Aor13HI, BseAI, Bsp13I, BspEI, MroI).
Invitrogen™ Anza™ 17 KpnI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Invitrogen™ Anza™ 29 KflI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Promega Restriction Enzymes - Kpn I Blue/White Cloning Qualified (J)
Capable of digesting DNA in 15 minutes or less.
Thermo Scientific™ Buffer KpnI Available on GSA/VA Contract for Federal Government customers only.
Optimize digestions with KpnI restriction enzyme using this buffer, premixed with BSA for enhanced stability.
Invitrogen™ Anza™ 60 Kpn2I Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Invitrogen™ Anza™ 50 KspAI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
  spinner