Extraction Systems
Wheaton™ Three-ball Snyder Column Available on GSA/VA Contract for Federal Government customers only.
For concentration of trace amounts of analyte in organic solvent
PYREX™ Soxhlet Apparatus Allihn Condenser Extraction Tube Available on GSA/VA Contract for Federal Government customers only.
Bulb in siphon tube near the top of tube facilitates cycling of the siphoning action
Kimble™ Kontes™ Kuderna-Danish Apparatus with a Graduated Concentrator Tube
The Kuderna-Danish apparatus was developed for the concentration of trace amounts of sample dissolved in organic solvents. The column is designed to speed evaporation with reduced hold-up. It is very useful in sample preparation before analysis with solvents such as petroleum ether or hexane.
Kimble™ Kimble&trade Kontes™ Extractor Body for Heavier Than Water Continuous Liquid Extraction Apparatus
Replacement component of 584190-0000, this Extractor Body designed for EPA priority samples where the extracting solvent is heavier than water.
PYREX™ Kuderna-Danish Replacement Flasks Available on GSA/VA Contract for Federal Government customers only.
Includes pair of 1.25 in. stainless-steel springs and two sizes of Keck clamps
Kimble™ Kontes™ 25mL Concentrator Tube for Evaporative Concentrators
This concentrator tube, for use with Kuderna-Danish and other concentrators, may be detached from the complete assembly and stoppered for analysis or storage.
Whatman™ Grade 603 Cellulose Extraction Thimbles
Range in size and length to ensure optimal fit.. Whatman™ Grade 603 Cellulose Extraction Thimbles are for use with extracting pesticides from environmental samples and fat analysis in the food industry
Kimble™ Kontes™ 10mL Tapered Concentrator Tube for Evaporative Concentrators Available on GSA/VA Contract for Federal Government customers only.
This tapered concentrator tube, for use with Kuderna-Danish and other concentrators, may be detached from the complete assembly and stoppered for analysis or storage.
Whatman™ Cellulose Soxhlet Extraction Thimbles, single 1mm wall
Manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Whatman™ Cellulose Soxhlet Extraction Thimbles are intended for applications including use in air and waste gas analysis.
Kimble™ Kontes™ Concentrator Tubes for Evaporative Concentrators
Used with Kontes Evaporative Concentrators.
Labconco™ Goldfisch Fat Extractor Accessory, Beaker
Accessory part for use with Labconco Goldfisch Fat Extractors.
PYREX™ Accelerated One-Step™ Extractor/Concentrator Replacement Parts Available on GSA/VA Contract for Federal Government customers only.
Replacement parts for Accelerated One-Step Extractor/Concentrator Systems
Whatman™ High Purity Glass Microfiber Soxhlet Extraction Thimbles
Designed to fit most Soxhlet extraction units. Whatman™ High Purity Glass Microfiber Soxhlet Extraction Thimbles are made of 100% borosilicate glass microfiber. They are well-suited to pollution monitoring when analysis of hot and acidic gases is required.
Whatman™ Cellulose Soxhlet Extraction Thimbles, double 2mm wall
Manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Whatman™ Cellulose Soxhlet Extraction Thimbles are intended for applications including use in air and waste gas analysis.
Foss™ Accessories for Soxtec™ Fat Extraction Systems: Cup Holder and Stand
For use with Soxtec™ 2055 system and discontinued systems
Kimble™ Kontes™ Soxhlet Extraction Body only, ST 45/50 24/40
Extractor body only, used as component part of 585050 Soxhlet extraction assemblies.
Wheaton™ 190mm Soxhlet Extractor With 24/40 Bottom Inner Joint and 34/45 Top Outer Joint
For continuous extraction of analytes from solid into organic solvent
Kemtech America™ Synthware™ Soxhlet Extractor
For the extraction of solids with a suitable solvent
  spinner