Moisture Meters
Fisherbrand™ Specific Gravity Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Double scales can be read without turning the hydrometer.
Fisherbrand™ Traceable™ Digital Hygrometer/Thermometer
Hang on the wall or use included plastic stand; housed in rugged ABS case
Fisherbrand™ Baume Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Baume “heavy” scale for working with liquids heavier than water.
Fisherbrand™ Set of Eight Precision Specific Gravity Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Specific Gravity Hydrometers are permanently marked with individual serial numbers.
Fisherbrand™ Midget Urinometer
Urinometer accuracy guaranteed to ±0.002.
Fisherbrand™ Specific Gravity/Baume Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Fisherbrand™ Specific Gravity/Baume Hydrometers are calibrated according to ASTM™ E 126 specifications.
H-B Instrument™ Durac™ Baume Plastic Plain Form Hydrometers
Safety and quality protection in one instrument!
Fisherbrand™ Alcohol Hydrometers Available on GSA/VA Contract for Federal Government customers only.
Used by distillers, law enforcement and taxing agencies to measure the alcoholic content of beverages.
Fisherbrand™ Soil Analysis ASTM Hydrometers
These hydrometers are used to determine the specific gravity of soil in the range of 0.995 to 1.038 or -5 to 60g/L (using a weight by volume scale).
Fisherbrand™ Baume Plain Form Hydrometers
Available in a variety of models measuring Baume of syrups and other liquids heavier than water in various ranges from 0° to 70°
Fisherbrand™ ASTM™ Hydrometers, Plain Form
API hydrometers are available in regular and ASTM™ recommended forms.
H-B Instrument™ DURAC™ Safety Precision Combined Form Brix Sugar Hydrometers
These combined-form hydrometers are used to measure % sucrose by weight in various ranges from 0° to 101° Brix, and include a convenient, built-in thermometer with a temperature scale of 0/50°C
Fisherbrand™ Salimeters Available on GSA/VA Contract for Federal Government customers only.
Fisherbrand™ Salimeters are used to measure pickling and meat-packing brines.
H-B Instrument™ Durac™ Salt Brine Plain Form Hydrometers
Choose from two plain form models with ranges from 0 to 100% or 0 to 26.5% to measure sodium chloride saturation of salt brines
Fisherbrand™ Replacement Hydrometer for Midget Urinometer
For use with Fisher Scientific Midget Urinometer (cat. no. 22274209).
Fisherbrand™ Replacement Hydrometer for Squibb Urinometer
For use with Fisherbrand™ Squibb Urinometer (cat. no. 22274233).
H-B Instrument™ Durac™ API ASTM Hydrometers; Traceable to NIST
Measure the relative density of oil and petroleum products in ranges from -1° to 101° API with these hydrometers. H-B Instrument Durac™ API ASTM Hydrometers conform to American Petroleum Institute (API) and the American Society of Testing Materials (ASTM) E100 standards. Traceable to NIST.
H-B Instrument™ Durac™ API ASTM Hydrometers with Individual Calibration Report; Traceable to NIST
Measure the relative density of oil and petroleum products in ranges from -1° to 101° API with these hydrometers. H-B Instrument Durac™ API ASTM Hydrometers conform to American Petroleum Institute (API) and the American Society of Testing Materials (ASTM) E100 standards. Traceable to NIST, with Individual Calibration Report.
Fisherbrand™ Plain Form Petroleum Hydrometers Available on GSA/VA Contract for Federal Government customers only.
API Plain Form Petroleum Hydrometers are available in regular and ASTM™ recommended forms.
H-B Instrument™ Durac™ Specific Gravity/Relative Density (g/cm&superscript_3;) Plain Form Hydrometers: for Liquids Heavier Than Water
Available in a variety of models measuring specific gravity/relative density (g/cm&superscript_3;) of liquids heavier than water in various ranges.
Fisherbrand™ ASTM™ Specific Gravity Hydrometers for Liquids Lighter than Water
Accuracy of hydrometer meets or exceeds ASTM™ E 100 specifications.
Fisherbrand™ Safety Blue Thermohydrometer, Fahrenheit Available on GSA/VA Contract for Federal Government customers only.
Lead and mercury free thermohydrometers.
DWK Life Sciences Kimble Chase™ Precision Specific Gravity Hydrometers
Available in a variety of graduation ranges and overall lengths
Fisherbrand™ Brix Hydrometers
Used to measure the concentration of sugar solutions in refineries.
  spinner