Refractometers
Mettler Toledo™ Refracto™ 30GS Portable Refractometer Available on GSA/VA Contract for Federal Government customers only.
Offers an extended measuring range due to its special prism design for measurements in laboratory and production environments.
Fisherbrand™ Handheld Analog Clinical Refractometer
When you need reliable, accurate readings of your Urine samples, the Fisherbrand™ Analog Clinical Refractometers delivers the value with the ability to read Serum Protein, Specific Gravity and Refractive Index all on a single scale.
Fisherbrand™ Handheld Analog Salinity Refractometer
When you need reliable, accurate readings of your salt water, the Fisherbrand™ Analog Salinity Refractometers delivers the value with both readings of Salinity and Specific Gravity all in one scale.
Laxco™ Benchtop Digital Brix/RI Refractometer
Laxco™ Benchtop Digital Brix Refractometers brings measuring of Brix & RI to a high degree of accuracy, with the ability through its GIBO software to track, measure, graph, & export data.
Fisherbrand™ Handheld Analog Brix/Sucrose Refractometer
When you need reliable, accurate readings of your sugar or sucrose levels, the Fisherbrand™ Analog Brrix Refractometers delivers value with a range of brix options
Laxco™ Benchtop Digital Salinity Refractometer
Laxco™ Benchtop Digital Salinity Refractometers brings measuring of NaCi and Seawater Salinity to a high degree of accuracy, with the ability through its GIBO software to track, measure, graph, & export data.
Reichert™ NIST Certified Calibration Oil
Check the accuracy of an instrument using calibration oils. Reichert™ NIST Certified Calibration Oil are formulated laser liquid calibration oils that are supplied with an NIST certification of traceability.
Cargille Laboratories Refractive Index Liquid Reference Standards
Choose from seven single sets of Refractive Index liquids
Laxco™ 60° Brix Calibration Fluid for Handheld Analog Refractometers
Used to Calibrate Handheld Analog Brix Refractometers
Reichert™ Handheld Digital Refractometers
Big features and performance in a small package
Laxco™ 46° Brix Calibration Fluid for Handheld Analog Refractometers
Used to Calibrate Handheld Analog Brix Refractometers
Laxco™ Replacement Daylight Plate for Handheld Analog Refractometer
Pack of 5 Replacement Daylight Plates for Laxco™ for Handheld Analog Refractometer
Reichert™ AR200™ Handheld Digital Refractometer
Lightweight digital refractometer designed for the food, beverage, agricultural, industrial-fluid, chemical, petrochemical, pharmaceutical, medical, and plastic industries
Reichert™ Refractometer Table Stand: Replacement Bulb
Fits table stand for Goldberg handheld refractometers.
Reichert™ Reichert™ Rhino Ultra-Durable Handheld Refractometers
Designed specifically for rugged factory and field use—fully IIP67 water rresistant, dustproof, and impact resistant
Laxco™ Handheld Analog Brix/Sucrose Refractometer
When you need reliable, accurate readings of your sugar or sucrose levels, the Laxco™ Analog Industrial Brrix Refractometers rugged design is built to last. A wide range of brix options makes it the ideal choice.
Reichert™ Salt-Chek Digital Handheld Refractometer
Automatic measurements with simple three button operation (Read, scale and cal.)
Fisherbrand™ Handheld Analog Automotive Refractometer
When you need reliable, accurate readings of your Battery Fluid or Antifreeze, the Fisherbrand™ Analog Automotive Refractometers delivers the value with ability to ready Ethylene, Propylene, and Battery Fluid measurements on one scale.
Reichert™ EG-Chek Digital Handheld Refractometer
Meets ASTM D3321-12 Standard test method for field test determination for freezing point of aqueous engine coolants
Reichert™ Rhino Clinical Handheld VET360 Refractometer
Rugged portable refractometer for urine specific gravity and serum/plasma protein readings
Reichert™ USG-Chek Digital Handheld Refractometer
Urine specific gravity refractometer suited for medical clinic
Laxco™ Handheld Analog Salinity Refractometer
When you need reliable, accurate readings of your salt water, the Laxco™ Analog Industrial Salinity Refractometers rugged design make is built to last. A single scale allows for the ability to read both Salinity and Specific Gravity all in one scale, making it the ideal choice.
Laxco™ Handheld Analog Clinical Refractometer
When you need reliable, accurate readings of your Urine samples, the Laxco™ Analog Industrial Clinical Refractometers rugged design is built to last. With the ability to read Serum Protein, Specific Gravity and Refractive Index all on a single scale, it makes it the ideal choice.
Fisherbrand™ Analog Abbe Benchtop RI Refractometer PROMO
FisherBrand™ Analog Abbe Benchtop RI Refractometer uses a digital thermometer to track temperature fluctuations throughout the sample reading processes to ensure an accurate reading each and every time.
Fisherbrand™ Analog Abbe Benchtop RI Refractometer
FisherBrand™ Analog Abbe Benchtop RI Refractometer uses a digital thermometer to track temperature fluctuations throughout the sample reading processes to ensure an accurate reading each and every time.
Fisherbrand™ Handheld Analog Refractometer Daylight Plate Replacement Assembly
Replacement Daylight Plate with Collar for all Fisherbrand™ Handheld Analog Refractometer
Mettler Toledo™ Excellence RM Digital Benchtop Refractometers Available on GSA/VA Contract for Federal Government customers only.
Benchtop refractometers allow you to determine the refractive index, BRIX, HFCS, or user-defined concentrations of a wide variety of liquids quickly and accurately. Mettler Toledo Excellence RM Digital Benchtop Refractometer has an in-built Peltier temperature control which provides both reliable temperature stability and rapid heating/cooling rates.
Mettler Toledo™ Refracto™ 30PX Portable Refractometer Available on GSA/VA Contract for Federal Government customers only.
For measurements in laboratory and production environments.
  spinner