Thermo Scientific Liquid Handling Solutions
Featured Products
With a history of innovation, the Thermo Scientific™ portfolio continues to offer breakthrough technology.
Browse Thermo Scientific Liquid Handling Products:
My Pipette Creator App
Use the My Pipette Creator app to manage and share pipette programs from the comfort of your computer. Save time, reduce errors, and increase accuracy and reproducibility when you send programs to your Thermo Scientific™ E1-ClipTip™ Pipettes. Create programs or download pre-programmed protocols for specific qPCR and sample prep assay kits, then transfer the programs directly to your pipette via Bluetooth or micro-USB.
Learn More
F1-ClipTip Pipetting System
The new Thermo Scientific™ F1-ClipTip™ Manual Pipetting System features innovative, interlocking technology that allows you to feel the tips lock firmly in place.
Thermo Scientific F1-ClipTip Manual Pipetting System
Ensure reproducible results and a secure seal on every channel with the Thermo Scientific™ F1-ClipTip™ Manual Pipetting System. Minimal force is needed for tip attachment and ejection. Compatible with Thermo Scientific™ ClipTip™ Pipette Tips.
Thermo Scientific Finnpipette F1 Pipetting System
The Thermo Scientific™ Finnpipette™ F1 Pipetting System is remarkably lightweight and designed with increased air boost for efficient delivery of microvolumes. For best results, use with Thermo Scientific™ Finntip™ Pipette Tips.
Thermo Scientific Finnpipette F2 Pipetting System
For accurate microvolume dispensing, choose the Thermo Scientific Finnpipette F2 Pipetting System. These pipettes are designed with a large volume display and are fully autoclavable. For best results, use with Thermo Scientific™ Finntip™ or Finntip™ Flex™ Pipette Tips.
Electronic Pipetting Systems
Electronic Pipettes and Tip Systems help blend the control and lightweight design of manual pipettes with the precision and accuracy of electronic pipettes to boost your productivity.
E1-ClipTip Electronic Pipettes enable efficient, centralized programming and sharing of protocols between pipettes and colleagues, including access to pre-programmed protocols for many of your favorite reagent kits — all from the comfort of your computer. To learn how to connect your pipette to the My Pipette Creator App, click here and watch the video titled “Thermo Scientific My Pipette Creator” under More Resources.
Watch the video to the right to explore the features of Finnpipette Novus Electronic Pipettes.
E1 ClipTip Electronic Equalizer Pipettes
Meet Your New Favorite Pipette
Perform sample transfers between virtually any tube, rack, microplate, or horizontal gel box quickly and efficiently with the Thermo Scientific E1-ClipTip Equalizer Pipette. Adjustable tip spacing allows you to set the distance between tips by simply sliding the scale to expand or contract to the desired labware format. See it in action above.
Thermo Scientific E1-ClipTip Electronic Pipettes
The Thermo Scientific E1-ClipTip Pipetting System features a unique interlocking design for secure seals, index finger pipetting, and electronic tip ejection. For best results, use with Thermo Scientific ClipTip Pipette Tips.
Thermo Scientific Finnpipette Novus Electronic Pipettes
The Thermo Scientific Finnpipette Novus Electronic Pipetting System offers single-finger pipetting operation in single and multichannel models. For best results, use with Finntip or Finntip Flex Tips.
Manual and Robotic Pipette Tips
Improving Pipette Performance and Results
Choose from a wide range of features to find the best pipette tips for your needs, including low retention, extended length, wide bore, and gel loading. The Thermo Scientific brand has tips to fit nearly every pipette and automated liquid handler.
ART Pipette Tips Video
Count on the performance and reliability of Thermo Scientific™ ART™ barrier, non-filtered, and specialty tips for any application and pipette. Enjoy the complete universal tip solution with zero contamination and versatile storage along with space-saving reload towers.
Thermo Scientific ART Pipette Tips
The Thermo Scientific™ ART™ Pipette Tip line includes barrier and non-filtered tips from 10μL to 10mL, as well as gel-loading and wide-bore specialty products. ART tips have a self-sealing barrier that guarantees 100% protection against aerosol and liquid contamination. Every ART tip is certified RNase, DNase, ATP, and pyrogen free.
Thermo Scientific ART SoftFit L Pipette Tips
Thermo Scientific™ ART™ SoftFit L™ Pipette Tips provide a complete selection of barrier, non-filtered, and specialty tips. These tips are available in standard and low-retention options and designed for use with Rainin LTS pipettes.
Thermo Scientific Robotic Tips
Designed to fit over 50 workstations, these robotic tips undergo rigorous quality control testing and have a low coefficient of variation (from 1 to 3%).
Thermo Scientific Good Laboratory Pipetting Practices Brochure
Find information about tools and techniques that can help improve the accuracy and precision of your pipetting.
Thermo Scientific ART Pipette Tips Brochure
The Thermo Scientific ART portfolio offers a complete selection of barrier, non-filtered, and specialty tips. Learn more about Thermo Scientific ART Pipette Tips.
Thermo Scientific ClipTip Pipetting Systems Brochure
Learn about worry-free pipetting with the Thermo Scientific ClipTip Pipetting System. Read about the features of F1-ClipTip Pipettes, E1-ClipTip Pipettes, and more.
Thermo Scientific Finnpipette Manual Pipetting Systems Brochure
Learn about the features and specifications of Thermo Scientific Manual Pipetting Systems, including Finnpipette F1, Finnpipette F2, Finntip and Finntip Flex Pipette Tips, and more.
More Resources
Thermo Scientific Finnpipette Novus Electronic Pipette Brochure
Thermo Scientific Finnpipette Novus Single and Multichannel Electronic Pipettes deliver exceptional performance. Read about the intuitive programming, index-finger operation and more.
Thermo Scientific Versette Automated Liquid Handler Brochure
Read about the Thermo Scientific Versette user-friendly liquid handler. This liquid handling solution offers a choice of 96- or 384-chanel pipetting heads.
Thermo Scientific Automation Tips and Accessories Brochure
Browse the Thermo Scientific Automation Tips and Accessories brochure to read about available tips, workstations, and more.
Thermo Scientific My Pipette Creator
Boost your productivity and research results by pairing your Thermo Scientific E1-ClipTip Pipettes with the Thermo Scientific My Pipette Creator App.
ART Pipette Tips Video
Count on the performance and reliability of Thermo Scientific™ ART™ barrier, non-filtered, and specialty tips for any application and pipette. Enjoy the complete universal tip solution with zero contamination and versatile storage along with space-saving reload towers.
Consistent, Reproducible Pipetting
Experience a breakthrough in technology that makes daily pipetting less of a chore with the Thermo Scientific™ ClipTip™ Pipetting System — it starts with interlocking pipette tips that seal in security.
Manual or electronic Thermo Scientific ClipTip Pipettes are available with fixed or adjustable volumes and in single or multichannel configurations.
Simply fill out the form to request a demonstration.