Thermo Scientific MaxQ Shakers
max-q-shakers-d
Featured Products
Trust your samples to the outstanding line of Thermo Scientific™ MaxQ™ shakers.
MaxQ basic and advanced shakers contain a triple-eccentric drive mechanism to accommodate heavier loads and provide uniform shaking. Designed to ensure 24/7 operation, MaxQ shakers also feature maintenance-free brushless DC motors.
  • 15 to 500rpm speed range
  • View temperature, speed and time simultaneously on three individual LED displays
  • Continuous timed operation from 0.1 hour to 999 hours or 0.1 minute to 999 minutes
  • Audible/visual alarms protect cultures from temperature and speed deviations
Top Product Categories:
Open Air
Get the choice and flexibility you need from a variety of basic benchtop shaker models. Separately available platforms for some models allow you to increase capacity simply by changing the platform.
basic-maxq-3000-2000
MaxQ 2000 and 3000
• Small shaker footprint saves valuable bench space 
• Units operate in temperature range of 0°C to +40°C and humidity conditions from 20 to 80% non-condensing, enabling them to be used in a variety of environments
• Triple-eccentric drives handle heavy platform loads (MaxQ 2000: 35 lb., MaxQ 3000: 50 lb.)
Browse Now
thermo-sci-max-q-2506
MaxQ 2506
• Analog operating system is affordably priced
• Interchangeable 13 x 11", 18 x 18" and 24 x 18" platforms are available to accommodate a variety of labware
• On dual action models, control lever changes shaking motion while unit is stationary or in operation
Browse Now
thermo-sci-max-q-2508
MaxQ 2508
• Variable speed control allows gentle to vigorous agitation
• Built-in timer facilitates repeatable results
• Control lever changes the motion while the shaker is stationary or in operation
Browse Now
Incubated/Refrigerated
Upgrade to an advanced shaker for your large-capacity applications. Select the right shaker for your needs and customize by selecting either a universal platform or a dedicated platform.
advanced-maxq-4000
MaxQ 4000
• Large chamber holds six 2L flasks
• Clear acrylic lid provides high visibility for sample harvesting
• Safety interlock protects operator by stopping platform motion when lid is lifted 
Browse Now
thermo-sci-max-q-4450
MaxQ 4450
• Space-saving shaker designed for a small footprint
• Holds up to four 1L flasks
• Requires minimal bench space 
Browse Now
thermo-science-max-q-5000
MaxQ 5000
• Floor model
• Large chamber holds four 6L flasks
• Easy-access drain system with quick-disconnect fitting permits easy clean up of spills
• Foot pedal opens lid for hands-free operation 
Browse Now
thermo-science-max-q-7000
MaxQ 7000
• Easy-access drain with quick disconnect located in the front of the cabinet for convenient draining and disinfecting
• Alarm alerts operator in case of low water level
• Optional cooling coil available for near-ambient temperatures and below
Browse Now
Stackable
Maximize flexibility and production capability with stackable shakers. Select the right shaker for your needs and customize by selecting either a universal platform or a dedicated platform.
stackable-maxq-6000
MaxQ 6000
• Units can be stacked two high on the floor to save valuable bench space
• Two adjustable-height shelves provide added storage
• Large viewing window and internal light offer sample visibility 
• Includes stacking kit 
Browse Now
stackable-maxq-8000
MaxQ 8000
• Units can be stacked two to three high for space savings with a speed range of 25 to 400rpm (±1rpm)
• HEPA filter evenly distributes air inside the chamber and reduces cross-contamination, improving culturing conditions; rated at 95% efficient at 0.3µm for entrapment of airborne particles
• Ergonomically designed fold-down door and a platform that slides out 100% with a maximum platform height of 61" (154.9cm) when stacked three units high improves sample handling 
• Includes stacking kit
Browse Now
High Performance
When you need a reliable solution for sensitive cell cultures requiring protection from contamination, choose from the MaxQ high-performance line of shakers.
  • 25 to 525rpm speed range
  • Backlit display contains run points and set points of temperature, speed and time that can be viewed simultaneously
  • Continuous operation or timed operation from 1 minute up to 199 hours and 59 minutes
  • Audible/visual alarms protect cultures from temperature and speed deviations
  • HEPA filter for maximum protection from contamination
  • Directed airflow system for optimum temperature uniformity and ideal growth conditions even when the units are full
high-performance-maxq-416
MaxQ 416 HP and 430 HP
• Low heat 24V DC motor with moisture-resistant electronics provides continuous maintenance-free operation on the benchtop or in a chamber
• Triple-counter-balanced mechanism for optimum handling of full or unbalanced loads, regardless of vessel placement; completely eliminates "walking"
• Units operate in temperature range of 4°C to +40°C and humidity conditions from 50 to 80% non-condensing enabling them to be used in a variety of environments 
Browse Now
thermo-scientific-hp-420
MaxQ 420 HP
• Twin blowers and gasketed lid ensure temperature uniformity of ±0.3°C (in flask), creating an ideal environment for temperature-sensitive cultures
• Triple-counter-balanced mechanism
• Microprocessor functions are calibrated through keypad
Browse Now
thermo-scientific-hp-435
MaxQ 435 HP, 436 HP and 481 HP
• Floor model
• HEPA filtration for maximum protection from contamination
• Drain system with valve makes it easy to eliminate liquid, reducing the risk of water-borne contamination
• Directed laminar airflow system for optimum temperature uniformity 
Browse Now
All Products
Filtered Search Results
Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.
Narrow Results
Narrow Results
  • (237)
  • (36)
  • (11)
  • (18)
  • (33)