The New Transferpette S : A Perfect Fit for Every Hand
Experience how simple, easy, and efficient pipetting can be with the new BRAND Transferpette S. Crafted with high-grade materials, precision workmanship, and the quality of German manufacturing, the Transferpette S line of air displacement pipettes offers a host of updated features:
- The grip has been designed for a better fit in left and right hands of all sizes
- All functions — volume setting and locking, pipetting, and tip ejection — can be comfortably operated single handedly without shifting position
- Low operating forces, a short pipetting stroke, and easy tip mounting and ejection ensure comfortable pipetting
- Volume setting is quick and easy with a four-digit volume display that is always visible, with an easy-to-operate thumbwheel to rapidly select the desired volume
- The thumb-operated volume change protection mechanism prevents inadvertent volume drift during pipetting for additional reliability and improved reproducibility
- Color-coded pipette accents simplify pipette and tip selection
- Single- and multi-channel models are available, as well as sets with volume ranges from 0.1 µL to 10 mL
- All Transferpette S pipettes are made of UV-resistant materials and can be repeatedly autoclaved at 121°C for decontamination
Single-channel models include a three-year warranty and feature a slimmer shaft that fits narrow vessels — including 15 mL conical tubes, housing designed for easy cleaning, and tip cones designed to fit most brands of quality pipette tips. The lower housing can be easily disassembled without tools for cleaning and maintenance. A special “sub-micro” model provides unsurpassed accuracy in the 0.1 to 1 µL range, and 10 adjustable-volume models cover the 0.1 µL to 10 mL range. Ten pipettes in five sizes cover the volume range from 0.5 to 300 µL. Eight different fixed-volume models are also available.
Multichannel pipettes are available in 8- and 12-channel models. The patented tip cone design allows the user to easily remove individual tip cones for cleaning or replacement in the lab, reducing downtime. The advanced O-ring design and stepped tip ejector improve tip fit and reduce tip mounting and ejection forces even further.
In addition to Transferpette S pipettes, BrandTech offers a full line of liquid handling products, including the new HandyStep touch repeating pipette. The HandyStep touch features a touchscreen interface with intuitive menu-driven operation and integrated help function. Operating modes include multi-dispensing, auto-dispensing, and pipetting. Automatic tip ejection simplifies use and additional modes — sequential dispensing, multi-aspiration, and titration — are also available.
![BrandTech logo_2c.Reg.FINAL [Converted] BrandTech logo_2c.Reg.FINAL [Converted]](/us/en/scientific-products/publications/lab-reporter/2020/issue-4/the-new-transferpette-s-a-perfect-fit-for-every-hand/jcr%3acontent/content-par-main/columncontrol_d5f1/colPar2/imageandrte_e208.img.png/1680531511292.png)