Chemicals and Consumables for PFAS Testing
PFAS Testing Products
Per- and polyfluorinated alkyl substances (PFAS) are manufactured chemicals found in common consumer products and used in a variety of industries. They are known as “forever chemicals” because they do not break down and can bioaccumulate. As you set out to test for these substances, find the products you need to complete each step.
Chemicals  |  Columns and Accessories  |  Sample Handling  |  Sample Preparation
Chemicals
Prepare your samples with the right solvents and reagents, including Thermo Scientific™ UHPLC grade solvents that meet all current EU, EPA, and ISO regulations.
Chemicals for Testing
    Catalog Number Description Price Quantity & Availability  
Success
A4581 Details
Methanol, UHPLC-MS Quantity: 1 L
Add to Cart
Product Specifications
Melting Point -98°C
Titratable Base 0.0002meq/g max.
Boiling Point 64.7°C
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
MDL Number MFCD00004595
Water 0.02% Max.
Titratable Acid 0.0003meq/g max.
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Assay Percent Range 99.9% minimum by GC
Vapor Pressure 128 hPa at 20°C
Viscosity 0.55 cP at 20°C
Grade UHPLC-MS
CAS 67-56-1
PubChem CID 887
Physical Form Liquid
Absorbance ≤0.005 AU at 280nm, ≤0.01 AU at 254nm, ≤0.1 AU at 230nm, ≤0.4 AU at 215nm, ≤000 5 AU at 260nm, ≤02. AU at 220nm, ≤05. AU at 210nm
Packaging Clear Borosilicate Glass Bottle
Residue after Evaporation 1ppm max.
Quantity 1 L
Density 0.791 g/cm3
Linear Formula CH3OH
Color Colorless
SMILES CO
Chemical Name or Material Methanol
Molecular Formula CH4O
ChEBI CHEBI:17790
A929-1 Details
GSA_VA
Acetone, Optima™, for HPLC and GC, Fisher Chemical™ Quantity: 1 L
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Melting Point -95°C
Titratable Base 0.0006mEq/g max.
Boiling Point 56°C
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
MDL Number MFCD00008765
Water 0.5% max.
Titratable Acid 0.0003mEq/g max.
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
Assay Percent Range 99.9% min by GC
Vapor Pressure 247mbar at 20°C
Viscosity 0.32 mPa/s at 20°C
Grade OPTIMA™
CAS 67-64-1
Refractive Index 1.3555 to 1.3580 (at 25°C)
PubChem CID 180
Absorbance 1.00 AU max. 330nm, 0.06 AU max. at 340nm, 0.010 AU max. 350nm, 0.01 AU max. (350 to 400nm)
Physical Form Liquid
Formula Weight 58.08
Decomposition Information >4°C
pH 7
Packaging Wide Neck Amber Bottle
Residue after Evaporation 1ppm max.
Quantity 1 L
Density 0.790g/cm³
Identification Pass Test
Purity Grade Notes High purity mobile phase for HPLC and/or extraction solvent for GC applications
Percent Purity ≥99.5%
Linear Formula CH3COCH3
Color Colorless
SMILES CC(C)=O
Chemical Name or Material Acetone
Molecular Formula C3H6O
ChEBI CHEBI:15347
UN Number 1090
W81 Details
Water, UHPLC-MS Quantity: 1 L
Add to Cart
Product Specifications
Melting Point 0°C
Residue after Evaporation ≤1ppm
Boiling Point 100°C
Quantity 1 L
Synonym dihydrogen oxide, dihydrogen monoxide
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Density 1.000 g/cm3
Color Colorless
IUPAC Name oxidane
SMILES O
Vapor Pressure 17.5mmHg at 20°C
Molecular Weight (g/mol) 18.015
Grade UHPLC-MS
Chemical Name or Material Water
CAS 7732-18-5
Molecular Formula H2O
ChEBI CHEBI:15377
Physical Form Liquid
PubChem CID 962
Absorbance ≤0.005 AU at 254nm, ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 210nm, ≤0.01 AU at 220nm, ≤0.01 AU at 230nm, ≤0.01 AU at 240nm
pH 7
Packaging Clear Borosilicate Glass Bottle
P250-1 Details
Potassium Hydroxide (Pellets/Certified ACS), Fisher Chemical™ Quantity: 1 kg
Add to Cart
Product Specifications
Melting Point 360°C
Boiling Point 1320°C
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
MDL Number MFCD00003553
IUPAC Name potassium hydroxide
Assay Percent Range ≥85.0%
Molecular Weight (g/mol) 56.11
Grade Certified ACS
Assay Percent Notes ≥85.0%
CAS 1310-58-3
Physical Form Solid
PubChem CID 14797
Formula Weight 56.11
pH 13.5
Packaging Poly Bottle
Quantity 1 kg
Density 2.04g/cm³
Identification Pass Test
Percent Purity ≥85.0%
Color Yellow
SMILES [OH-].[K+]
Chemical Name or Material Potassium Hydroxide
Molecular Formula HKO
ChEBI CHEBI:32035
UN Number 1813
A11710X1-AMP Details
GSA_VA
Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™ Quantity: 10 x 1 mL
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Melting Point 8°C
Boiling Point 101°C
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
IUPAC Name formic acid
Vapor Pressure 44mbar at 20°C
Assay Percent Range ≥99.0 %
Molecular Weight (g/mol) 46.025
Grade OPTIMA™ LC-MS
CAS 64-18-6
Physical Form Liquid
Absorbance 0.01 AU max. at 254nm, 0.55 AU max. at 230nm, 0.85 AU max. at 220nm, 1.25 AU max. at 210nm
PubChem CID 284
Formula Weight 46.02
pH 2.1
Packaging Ampules
Residue after Evaporation 1ppm max.
Quantity 10 x 1 mL
Density 1.220g/cm³
Linear Formula HCOOH
Percent Purity ≥99.0%
Color Colorless
SMILES C(=O)O
Merck Index 15, 4269
Chemical Name or Material Formic Acid
Molecular Formula CH2O2
ChEBI CHEBI:30751
AC456870050 Details
Acetic acid, 50%, solution in water Quantity: 5 L
Add to Cart
Product Specifications
Quantity 5 L
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
MDL Number MFCD00036152
Flash Point >60°C
Percent Purity 50%
Linear Formula CH3CO2H
IUPAC Name acetic acid
SMILES CC(O)=O
Molecular Weight (g/mol) 60.05
Chemical Name or Material Acetic acid
CAS 64-19-7
Molecular Formula C2H4O2
ChEBI CHEBI:15366
Solubility Information Solubility in water: completely soluble.
PubChem CID 176
Formula Weight 60.05
AAA163430I Details
Ammonium acetate, 97% Quantity: 5000 g
Add to Cart
Product Specifications
Melting Point ∼114°C (decomposition)
Sensitivity Hygroscopic
Boiling Point (decomposition)
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
Quantity 5000 g
MDL Number MFCD00013066
Density 1.17
Flash Point 136°C (277°F)
Percent Purity 97%
Linear Formula CH3COONH4
IUPAC Name azanium;acetate
Beilstein 4186741
SMILES CC(=O)[O-].[NH4+]
Molecular Weight (g/mol) 77.083
Merck Index 14,495
Chemical Name or Material Ammonium acetate
CAS 631-61-8
Odor Ammonia-like
Molecular Formula C2H7NO2
ChEBI CHEBI:62947
Solubility Information Soluble in water,methanol,acetone and liquid ammonia.
PubChem CID 517165
Formula Weight 77.08
AAA1634330 Details
Ammonium acetate, 97% Quantity: 250 g
EDGE
Add to Cart
Product Specifications
Melting Point ∼114°C (decomposition)
Sensitivity Hygroscopic
Boiling Point (decomposition)
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
Quantity 250 g
MDL Number MFCD00013066
Density 1.17
Flash Point 136°C (277°F)
Percent Purity 97%
Linear Formula CH3COONH4
IUPAC Name azanium;acetate
Beilstein 4186741
SMILES CC(=O)[O-].[NH4+]
Molecular Weight (g/mol) 77.083
Merck Index 14,495
Chemical Name or Material Ammonium acetate
CAS 631-61-8
Odor Ammonia-like
Molecular Formula C2H7NO2
ChEBI CHEBI:62947
Solubility Information Soluble in water,methanol,acetone and liquid ammonia.
PubChem CID 517165
Formula Weight 77.08
AC270480010 Details
Formic acid, 99%, for analysis Quantity: 1 L
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Melting Point 8.0°C
Boiling Point 101.0°C
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
MDL Number MFCD00003297
IUPAC Name formic acid
Vapor Pressure 44mbar at 20°C
Assay Percent Range 99%
Molecular Weight (g/mol) 46.025
Grade Analytical
Refractive Index 1.3700 to 1.3720
CAS 64-18-6
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol, ether and glycerine
Physical Form Liquid
PubChem CID 284
Formula Weight 46.02
Packaging Plastic Bottle
Quantity 1 L
Density 1.2200g/mL
Flash Point 50°C
Specific Gravity 1.22
Fieser 01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177
Linear Formula HCO2H
Percent Purity 99%
Beilstein 02, 8
SMILES C(=O)O
Merck Index 15, 4269
Chemical Name or Material Formic acid
Molecular Formula CH2O2
ChEBI CHEBI:30751
Infrared Spectrum Authentic
AC326600025 Details
Dichloromethane, for residue and pestic. anal., stab. with amylene Quantity: 2.5 L
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Melting Point -97.0°C
Boiling Point 39.0°C to 40.0°C
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
MDL Number MFCD00000881
Water 0.02% max. (K.F.)
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
Vapor Pressure Vapor Pressure: 475hPa at 20°C
Viscosity 0.43 mPa.s (20°C)
CAS 75-09-2, 513-35-9
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
PubChem CID 6344
Physical Form Liquid
Formula Weight 84.93
Decomposition Information >120°C
Residue after Evaporation 2ppm max.
Quantity 2.5 L
Density 1.3250g/mL
Specific Gravity 1.325
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Linear Formula CH2Cl2
Beilstein 01, 60
SMILES ClCCl
Merck Index 15, 6135
Chemical Name or Material Dichloromethane
Molecular Formula CH2Cl2
ChEBI CHEBI:15767
Name Note ECD Tested, For Pesticide analysis
AA33285D9 Details
Ammonium hydroxide, ACS, 28.0-30.0% NH{3} Quantity: 5 x 1 kg
Add to Cart
Product Specifications
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
Quantity 5 x 1 kg
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
MDL Number MFCD00066650
Density 0.9
Color Colorless
IUPAC Name azanium;hydroxide
SMILES N.O
Molecular Weight (g/mol) 35.05
Assay Percent Range 28.0-30.0% NH3
Grade ACS
Chemical Name or Material Ammonium hydroxide
CAS 1336-21-6
Odor Ammonia-like
Molecular Formula H5NO
ChEBI CHEBI:18219
Solubility Information Miscible with water.
Physical Form Liquid
PubChem CID 14923
Formula Weight 35.05
AC423240010 Details
Acetone, 99.6%, ACS reagent Quantity: 1 L
Due to product restrictions, please to purchase or view availability for this product.
Product Specifications
Melting Point -95.0°C
Titratable Base 0.0006 meq/g max.
Boiling Point 56.0°C
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
MDL Number MFCD00008765
Water 0.5% max. (K.F.)
Titratable Acid 0.0003 meq/g max.
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
Assay Percent Range 99.6%
Vapor Pressure Vapor Pressure: 247mbar at 20°C
Viscosity 0.32 mPa.s (20°C)
Grade ACS Reagent
CAS Max % 100.0
CAS 67-64-1
Refractive Index 1.3585
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
PubChem CID 180
Physical Form Liquid
CAS Min % 99.5
Formula Weight 58.08
Decomposition Information >4°C
Packaging EcoSafPak™ Glass Bottle
Residue after Evaporation 0.001% max.
Quantity 1 L
Density 0.7900g/mL
Flash Point -20°C (-4°C)
Specific Gravity 0.79
Fieser 02,13; 03,4; 06,9
Percent Purity 99.6%
Linear Formula CH3COCH3
Beilstein 01, 635
SMILES CC(C)=O
Merck Index 15, 65
Chemical Name or Material Acetone
Molecular Formula C3H6O
ChEBI CHEBI:15347
UN Number 1090; Colorless
Columns and Accessories
Minimize contamination risk with the right products for your application, including the column types you see below.
Analytical, Guard, and GC Columns and Syringes
    Catalog Number Description Price Quantity & Availability  
Success
DX069689 Details
Thermo Scientific™ Acclaim™ 120 C18 Columns Length (Metric): 10 mm
Add to Cart
Product Specifications
Carbon Load 18%
Length (Metric) 10 mm
Quantity Each
Pore Size 120 Å
Particle Size 5 μm
Temperature 60°C
USP Type L1
Column Type Reversed Phase
Packing Material Ultrapure Silica
Diameter (Metric) 2.1 mm
Green Features Responsibly packaged
Product Line Acclaim
Stationary Phase C18
Column Format Guard Cartridge, 2 Pk.
Max. Pressure 4500 psi (310 bar)
Surface Area 300 m²/g
Phase Reversed Phase
Endcapped Yes
pH 2 to 8
26-096-1420 Details
GSA_VA
Thermo Scientific™ TraceGOLD TG-5SilMS GC Columns Length (Metric): 30 m
Add to Cart
Product Specifications
Description 0.25 um Thickness; 0.25 mm ID; 30 m Length
Quantity Each
For Use With (Application) GC/MS Applications Using Ion-Trap Systems, GC/MS for Polycyclic Aromatics, Phthalates, Phenols, Amines, and Organophosphates, GC/MS for Hydrocarbons Including Chlorinated Hydrocarbons
Length (Metric) 30 m
Particle Size 0.25 μm
USP Type G27, G36
Diameter (Metric) 0.25 mm
Max. Temperature 330/350°C
Stationary Phase TraceGOLD™ TG-5 SilMS
Product Line TraceGOLD
Diameter (Metric) Inner 0.25 mm
Polarity Low-Polar
Film Thickness 0.25 μm
17-626-012105 Details
Thermo Scientific™ Accucore™ RP-MS HPLC Columns Length (Metric): 10 mm
Add to Cart
Product Specifications
Carbon Load 7%
Length (Metric) 10 mm
Pore Size 80 Å
Particle Size 2.6 μm
For Use With Uniguard Direct-Connection and Stand Alone Holder
Particle Shape Spherical Solid Core
Temperature 60°C
USP Type N/A
Column Type Reversed Phase
Packing Material Spherical, Solid Core, Ultrapure Silica
Diameter (Metric) 2.1 mm
Product Line Accucore
Stationary Phase Solid Core Particles
Column Format Guard Cartridge, 4 Pk.
Max. Pressure 14500 psi (1000 bar)
Surface Area 130 m²/g
Phase Reversed Phase
Endcapped Yes
pH 2 to 9
03-151-659 Details
Thermo Scientific™ LinerGOLD™ GC Liners Description: LinerGOLD Splitless Liner, Single Taper; Quartz Wool
Add to Cart
Product Specifications
For Use With (Equipment) TRACE 1300 Series and Agilent Instruments
Product Type Splitless Liner with Single Taper
Description LinerGOLD Splitless Liner, Single Taper; Quartz Wool
Quantity 5
Length (Metric) 78.5 mm
Diameter (Metric) Inner 4 mm
Packing Material Quartz Wool
Diameter (Metric) Outer 6.3 mm
17-626-102130 Details
Thermo Scientific™ Accucore™ RP-MS HPLC Columns Length (Metric): 100 mm
Add to Cart
Product Specifications
Carbon Load 7%
Length (Metric) 100 mm
Quantity Each
Pore Size 80 Å
Particle Size 2.6 μm
For Use With HPLC
Particle Shape Spherical Solid Core
Temperature 60°C
USP Type N/A
Column Type Reversed Phase
Packing Material Spherical, Solid Core, Ultrapure Silica
Diameter (Metric) 2.1 mm
Product Line Accucore
Stationary Phase Proprietary
Column Format Analytical Column
Max. Pressure 14500 psi (1000 bar)
Surface Area 130 m²/g
Phase Reversed Phase
Endcapped Yes
pH 2 to 9
60180825 Details
GSA_VA
Thermo Scientific™ Super Clean™ Gas Cartridge Filters For Use With (Equipment): He
EDGE
Add to Cart
Product Specifications
For Use With (Equipment) He
Description Baseplate not included
Indicator Change Moisture: Yellow to Clear; Oxygen: Green to Gray
Type Indicating Triple Filter - Moisture, Oxygen, Hydrocarbons - He Preconditioned for GC/MS
Volume (Metric) 1.8 g H2O, 500 mL O2, 7 g HCs (as n-butane)
Technique GC/MS
Cartridge Type Triple
DX068982 Details
Thermo Scientific™ Acclaim™ 120 C18 Columns Length (Metric): 100 mm
Add to Cart
Product Specifications
Carbon Load 18%
Length (Metric) 100 mm
Quantity Each
Pore Size 120 Å
Particle Size 2.2 μm
For Use With LC/MS
Temperature 60°C
USP Type L1
Technique Bio-LC
Column Type Reversed Phase
Packing Material Spherical, Fully Porous, Ultrapure Silica
Diameter (Metric) 2.1 mm
Green Features Responsibly packaged
Product Line Acclaim
Stationary Phase C18
Column Format Analytical Column
Max. Pressure 11600 psi (800 bar)
Surface Area 300 m²/g
Phase Reversed Phase
Endcapped Yes
pH 2 to 8
01-213-149 Details
Thermo Scientific 10UL FN GT SMART SYRINGE
Add to Cart
DX069707 Details
Thermo Scientific™ Acclaim™ Guard Holder and Coupler For Use With (Equipment): All Acclaim Guard Cartridges
Add to Cart
Product Specifications
Green Features Responsibly packaged
For Use With (Equipment) All Acclaim Guard Cartridges
Includes Guard column holder and coupler
Product Line Acclaim
Description Acclaim Guard Cartridge Holder-Coupler Kit V-2
Quantity Each
Type Acclaim Guard Kit (Holder and Coupler) V-2
01-213-148 Details
Thermo Scientific 100UL FN GT SMART SYRINGE
Add to Cart
01-213-147 Details
Thermo Scientific 1ML FN GT SMART SYRINGE
Add to Cart
01-213-146 Details
Thermo Scientific 5ML RN GT SMART SYRINGE
Add to Cart
Sample Handling
Find the sample handling products you need to help ensure accurate results, even when PFAS concentrations are low.
Vials, Well Plates, and More
    Catalog Number Description Price Quantity & Availability  
Success
03-253-901 Details
Choice™ Nylon Syringe Filters Pore Size: 0.2 μm
Add to Cart
Product Specifications
Material (Membrane) Nylon
Product Line Choice
Quantity 100/Pk.
Prefilter No
For Use With (Application) HPLC, SPE
Pressure Rating 90 psi
Housing Material Solvent Resistent, Low-Extractable Polypropylene
Disposable Single-use
Pore Size 0.2 μm
Type Syringe Filter
Sterility Non-sterile
Diameter (Metric) 25 mm
03-376-232 Details
Thermo Scientific™ Target2™ PES (Polyethersulfone) Syringe Filters Pore Size: 0.2 μm
Add to Cart
Product Specifications
Quantity 100/Pk.
For Use With (Application) IC, SPE
Disposable Single-use
Pore Size 0.2 μm
Volume (Metric) <29 μL (Hold Up)
Type Syringe Filter
Sterility Non-sterile
Certifications/Compliance Target Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
Diameter (Metric) 17 mm
Diameter (Metric) Membrane 17 mm
Color Dark Gray
Material (Membrane) PES
Product Line Target2
Prefilter No
Housing Material Polypropylene
Pressure Rating 115 psi
Hold up Volume (After Air Purge) <35 μL
03-377-27 Details
GSA_VA
Thermo Scientific™ National Target All-Plastic Disposable Syringes Product Type: Luer-lock Syringe
Add to Cart
Product Specifications
For Use With (Equipment) Syringe Filter
Product Type Luer-lock Syringe
Product Line National Target
Description Luer-Lock Syringes; 3mL Capacity
Disposable Single-use
Volume (Metric) 3 mL
Sterility Non-sterile
Connection Type Luer Lock
Material Polyethylene (Plunger), Polypropylene (Barrel)
03-377-29 Details
GSA_VA
Thermo Scientific™ National Target All-Plastic Disposable Syringes Product Type: Luer-lock Syringe
Add to Cart
Product Specifications
For Use With (Equipment) SyringeFilter
Product Type Luer-lock Syringe
Product Line National Target
Description Luer-Lock Syringes; 10mL Capacity
Quantity Pack of 100
Disposable Single-use
Volume (Metric) 10 mL
Sterility Non-sterile
Connection Type Luer Lock
Material Polyethylene (Plunger), Polypropylene (Barrel)
03-377-28 Details
GSA_VA
Thermo Scientific™ National Target All-Plastic Disposable Syringes Product Type: Luer-lock Syringe
Add to Cart
Product Specifications
For Use With (Equipment) Syringe Filter
Product Type Luer-lock Syringe
Product Line National Target
Description Luer-Lock Syringes; 5mL Capacity
Disposable Single-use
Volume (Metric) 5 mL
Sterility Non-sterile
Connection Type Luer Lock
Material Polyethylene (Plunger), Polypropylene (Barrel)
03-377-23 Details
GSA_VA
Thermo Scientific™ National Target All-Plastic Disposable Syringes Product Type: Luer-slip Syringe
EDGE
Add to Cart
Product Specifications
For Use With (Equipment) Syringe Filter
Product Type Luer-slip Syringe
Product Line National Target
Description Luer-Slip Syringes; 10mL Capacity
Disposable Single-use
Volume (Metric) 10 mL
Sterility Non-sterile
Connection Type Luer Lock
Material Polyethylene (Plunger), Polypropylene (Barrel)
03-377-22 Details
GSA_VA
Thermo Scientific™ National Target All-Plastic Disposable Syringes Product Type: Luer-slip Syringe
EDGE
Add to Cart
Product Specifications
For Use With (Equipment) Syringe Filter
Product Type Luer-slip Syringe
Product Line National Target
Description Luer-Slip Syringes; 5mL Capacity
Disposable Single-use
Volume (Metric) 5 mL
Sterility Non-sterile
Connection Type Luer Lock
Material Polyethylene (Plunger), Polypropylene (Barrel)
03-377-24 Details
GSA_VA
Thermo Scientific™ National Target All-Plastic Disposable Syringes Product Type: Luer-slip Syringe
EDGE
Add to Cart
Product Specifications
For Use With (Equipment) Syringe Filter
Product Type Luer-slip Syringe
Product Line National Target
Description Luer-Slip Syringes; 20mL Capacity
Disposable Single-use
Volume (Metric) 20 mL
Sterility Non-sterile
Connection Type Luer Lock
Material Polyethylene (Plunger), Polypropylene (Barrel)
03-253-895 Details
Choice™ Nylon Syringe Filters Pore Size: 0.2 μm
Add to Cart
Product Specifications
Material (Membrane) Nylon
Product Line Choice
Quantity 100/Pk.
Prefilter No
For Use With (Application) HPLC, SPE
Pressure Rating 90 psi
Housing Material Solvent Resistent, Low-Extractable Polypropylene
Disposable Single-use
Pore Size 0.2 μm
Type Syringe Filter
Sterility Non-sterile
Diameter (Metric) 13 mm
03-377-21 Details
GSA_VA
Thermo Scientific™ National Target All-Plastic Disposable Syringes Product Type: Luer-slip Syringe
Add to Cart
Product Specifications
For Use With (Equipment) Syringe Filter
Product Type Luer-slip Syringe
Product Line National Target
Description Luer-Slip Syringes; 3mL Capacity
Disposable Single-use
Volume (Metric) 3 mL
Sterility Non-sterile
Connection Type Luer Lock
Material Polyethylene (Plunger), Polypropylene (Barrel)
03-253-890 Details
Choice™ PES (Polyethersulfone) Syringe Filters Pore Size: 0.2 μm
Add to Cart
Product Specifications
Material (Membrane) PES
Product Line Choice
Quantity 100/Pk.
Prefilter No
For Use With (Application) HPLC, SPE
Pressure Rating 90 psi
Housing Material Solvent Resistent, Low-Extractable Polypropylene
Disposable Single-use
Pore Size 0.2 μm
Type Syringe Filter
Sterility Non-sterile
Diameter (Metric) 25 mm
14-823-287 Details
Thermo Scientific™ Titan3™ Nylon Syringe Filters Prefilter: Yes
Add to Cart
Product Specifications
Quantity 100/Pk.
For Use With (Application) Sample Filtration: general use, most HPLC samples
Disposable Single-use
Pore Size 0.2 μm
Type Syringe Filter
Sterility Non-sterile
Certifications/Compliance Titan3 Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
Diameter (Metric) 30 mm
Diameter (Metric) Membrane 30 mm
Color Light Purple
Material (Membrane) Nylon
Product Line Titan3
Prefilter Yes
Housing Material Polypropylene
Pressure Rating 210 psi
Hold up Volume (After Air Purge) <157 μL
Color Code Purple
Sample Preparation
Extract and concentrate your PFAS samples to prepare them for screenings with minimal background contamination.
Cartridges, Filters, and Plastic Syringes
    Catalog Number Description Price Quantity & Availability  
Success
03-050-475 Details
Thermo Scientific™ Titan3™ PES (Polyethersulfone) Syringe Filters Pore Size: 0.2 μm
Add to Cart
Product Specifications
Quantity 200/Pk.
For Use With (Application) Sample Filtration: ion chromatography; tissue culture, proteins, nucleic acids, high-temperature liquids
Disposable Single-use
Pore Size 0.2 μm
Volume (Metric) <29 μL (Hold Up)
Type Syringe Filter
Sterility Non-sterile
Certifications/Compliance Titan3 Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520. Membrane: ICP Certified PES
Diameter (Metric) 17 mm
Diameter (Metric) Membrane 17 mm
Color Dark Gray
Material (Membrane) PES
Product Line Titan3
Prefilter No
Housing Material Polypropylene
Pressure Rating 115 psi
Hold up Volume (After Air Purge) <35 μL
03-376-222 Details
Thermo Scientific™ Target2™ PES (Polyethersulfone) Syringe Filters Pore Size: 0.2 μm
Add to Cart
Product Specifications
Quantity 100/Pk.
For Use With (Application) IC, SPE
Disposable Single-use
Pore Size 0.2 μm
Volume (Metric) <137 μL (Hold Up)
Type Syringe Filter
Sterility Non-sterile
Certifications/Compliance Target Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
Diameter (Metric) 30 mm
Diameter (Metric) Membrane 30 mm
Color Dark Gray
Material (Membrane) PES
Product Line Target2
Prefilter No
Housing Material Polypropylene
Pressure Rating 150 psi
Hold up Volume (After Air Purge) <157 μL
03-050-371 Details
Thermo Scientific™ Titan3™ Nylon Syringe Filters Prefilter: No
Add to Cart
Product Specifications
Quantity 200/Pk.
For Use With (Application) Sample Filtration: general use, most HPLC samples
Disposable Single-use
Pore Size 0.2 μm
Type Syringe Filter
Sterility Non-sterile
Certifications/Compliance Titan3 Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
Diameter (Metric) 17 mm
Diameter (Metric) Membrane 17 mm
Color Light Purple
Material (Membrane) Nylon
Product Line Titan3
Prefilter No
Housing Material Polypropylene
Pressure Rating 115 psi
Hold up Volume (After Air Purge) <35 μL
Color Code Purple
03-050-479 Details
Thermo Scientific™ Titan3™ PES (Polyethersulfone) Syringe Filters Pore Size: 0.2 μm
Add to Cart
Product Specifications
Quantity 100/Pk.
For Use With (Application) Sample Filtration: ion chromatography; tissue culture, proteins, nucleic acids, high-temperature liquids
Disposable Single-use
Pore Size 0.2 μm
Volume (Metric) <137 μL (Hold Up)
Type Syringe Filter
Sterility Non-sterile
Certifications/Compliance Titan3 Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520. Membrane: ICP Certified PES
Diameter (Metric) 30 mm
Diameter (Metric) Membrane 30 mm
Color Dark Gray
Material (Membrane) PES
Product Line Titan3
Prefilter Yes
Housing Material Polypropylene
Pressure Rating 210 psi
Hold up Volume (After Air Purge) <157 μL
03-253-901 Details
Choice™ Nylon Syringe Filters Pore Size: 0.2 μm
Add to Cart
Product Specifications
Material (Membrane) Nylon
Product Line Choice
Quantity 100/Pk.
Prefilter No
For Use With (Application) HPLC, SPE
Pressure Rating 90 psi
Housing Material Solvent Resistent, Low-Extractable Polypropylene
Disposable Single-use
Pore Size 0.2 μm
Type Syringe Filter
Sterility Non-sterile
Diameter (Metric) 25 mm
03-376-232 Details
Thermo Scientific™ Target2™ PES (Polyethersulfone) Syringe Filters Pore Size: 0.2 μm
Add to Cart
Product Specifications
Quantity 100/Pk.
For Use With (Application) IC, SPE
Disposable Single-use
Pore Size 0.2 μm
Volume (Metric) <29 μL (Hold Up)
Type Syringe Filter
Sterility Non-sterile
Certifications/Compliance Target Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
Diameter (Metric) 17 mm
Diameter (Metric) Membrane 17 mm
Color Dark Gray
Material (Membrane) PES
Product Line Target2
Prefilter No
Housing Material Polypropylene
Pressure Rating 115 psi
Hold up Volume (After Air Purge) <35 μL
03-377-27 Details
GSA_VA
Thermo Scientific™ National Target All-Plastic Disposable Syringes Product Type: Luer-lock Syringe
Add to Cart
Product Specifications
For Use With (Equipment) Syringe Filter
Product Type Luer-lock Syringe
Product Line National Target
Description Luer-Lock Syringes; 3mL Capacity
Disposable Single-use
Volume (Metric) 3 mL
Sterility Non-sterile
Connection Type Luer Lock
Material Polyethylene (Plunger), Polypropylene (Barrel)
03-377-29 Details
GSA_VA
Thermo Scientific™ National Target All-Plastic Disposable Syringes Product Type: Luer-lock Syringe
Add to Cart
Product Specifications
For Use With (Equipment) SyringeFilter
Product Type Luer-lock Syringe
Product Line National Target
Description Luer-Lock Syringes; 10mL Capacity
Quantity Pack of 100
Disposable Single-use
Volume (Metric) 10 mL
Sterility Non-sterile
Connection Type Luer Lock
Material Polyethylene (Plunger), Polypropylene (Barrel)
03-391-1B Details
Thermo Scientific™ Target2™ Nylon Syringe Filters Prefilter: No
Add to Cart
Product Specifications
Quantity 100/Pk.
For Use With (Application) HPLC, SPE
Disposable Single-use
Pore Size 0.25 μm
Type Syringe Filter
Sterility Non-sterile
Certifications/Compliance Target Polypropylene Syringe Filter Housings Meet The Requirements of 21 CFR 177.1520.
Diameter (Metric) 30 mm
Diameter (Metric) Membrane 30 mm
Color Pink
Material (Membrane) Nylon
Product Line Target2
Prefilter No
Housing Material Polypropylene
Pressure Rating 150 psi
Hold up Volume (After Air Purge) <157 μL
03-377-28 Details
GSA_VA
Thermo Scientific™ National Target All-Plastic Disposable Syringes Product Type: Luer-lock Syringe
Add to Cart
Product Specifications
For Use With (Equipment) Syringe Filter
Product Type Luer-lock Syringe
Product Line National Target
Description Luer-Lock Syringes; 5mL Capacity
Disposable Single-use
Volume (Metric) 5 mL
Sterility Non-sterile
Connection Type Luer Lock
Material Polyethylene (Plunger), Polypropylene (Barrel)
03-377-23 Details
GSA_VA
Thermo Scientific™ National Target All-Plastic Disposable Syringes Product Type: Luer-slip Syringe
EDGE
Add to Cart
Product Specifications
For Use With (Equipment) Syringe Filter
Product Type Luer-slip Syringe
Product Line National Target
Description Luer-Slip Syringes; 10mL Capacity
Disposable Single-use
Volume (Metric) 10 mL
Sterility Non-sterile
Connection Type Luer Lock
Material Polyethylene (Plunger), Polypropylene (Barrel)
03-377-22 Details
GSA_VA
Thermo Scientific™ National Target All-Plastic Disposable Syringes Product Type: Luer-slip Syringe
EDGE
Add to Cart
Product Specifications
For Use With (Equipment) Syringe Filter
Product Type Luer-slip Syringe
Product Line National Target
Description Luer-Slip Syringes; 5mL Capacity
Disposable Single-use
Volume (Metric) 5 mL
Sterility Non-sterile
Connection Type Luer Lock
Material Polyethylene (Plunger), Polypropylene (Barrel)
Featured Resources